Ward's Biology 2016-2017 - page 382
NervousSystem/LymphaticSystem
|
Models
382
SOMSO
®
Lymphatic
SystemModel
RemarkablyDetailed
Representation
ofComplexSystem
The intricatedepictionofthe lym-
phaticsystemclearlyshowsthepaths
bywhichfluidreturnsfromtissuesto
theblood-stream.Two-thirds life-size
andsotranslucent it looksreal,this
high-qualitymodelcanbeusedforall
levelsof instruction,from introductory
biologytoadvancedanatomy,to illus-
tratethe importanceofthesystem in
producingan immuneresponse.The
lymphaticcapillaries,ducts,andnodes
canallbeclearlyseen.Themodel is
mountedonabaseandcomeswitha
manualcontainingafold-outdiagram
and identifying30structures.Size:21¼"Lx33"Wx4¾"H.
$1,990.00
SOMSO
®
Nervous
System
Model
Half-Life-Size
FigureShows
EntireSystem
Clearlyviewdetailsofthe
centralandperipheral
nervoussystemwitha
modelofthewholehu-
manbody.Thesectioned
headshowsthebrain
andpartoftheskull;
thespinalnervesare
shown inrelationtothe
skeleton,themajorparts
ofwhicharealsofeatured
insection.Armsand legs
arepositionedtoshow
frontaland lateralelementsofthenervoussystem.
Themodel ismountedonabaseandcomeswitha
studyguide identifying33structures.Size:33¾"Lx
12½"Wx2¼"H.
$485.00
SOMSO
®
Central
Nervous
System
Model
Detailed,Accurate
Representation
Perfectfor
AdvancedStudy
Presentthehuman
nervoussystemwithan
incrediblyrealisticand
accurate life-sizemodel
oftheentirespinal
cord,fromthebrain
stemtothecoccygeal
plexus.Theventral
dissectionshowsall
nervebranches,and
sympatheticnervesare
shownonthe left,withtheirconnectionstothecentral
nervoussystem.Themodel ismountedonabaseand
includesamanual identifying50numberedstructures.
Size:12½"Lx35½"Wx7½"H.
$1,825.00
Full-FigureNervous
SystemPremiumModel
NerveEndingsareClearlyRevealed
Discoveraheadtotoeviewofthehuman
nervoussystembeneaththeouter-bodysurface
withaccuraterepresentation.The lefthalfofthe
skullhasbeenremovedtorevealthebrainand
medulla,andthespineandpelvishavebeen
cutverticallytoshowacross-sectionalview.
Inthestomach,distributionofthevagusand
sympatheticnervesarerevealed.Terminationof
nerveendings isfullyshown.Modelheight is50",
and ismountedonametalstand. Includeskey.
FordistributionwithinNorthAmericaonly.
$3,400.00
3B
®
Nervous
System
Model
Half-Life-Size
FigureShows
EntireSystem
Viewthecentraland
peripheralnervous
systemswiththis
detailedreliefmodel.
It includesakey iden-
tifying47structures.
Size:31½"Hx13"Wx
2¼"D.
$425.00
NervousSystem LymphaticSystem
NervousSystemActivityModel
OverviewofStructurefromSeveralDifferentPerspectives
Featuringthreemodels inone,theraised-reliefplaquedepictsthehumannervoussystem inanumberof
differentways.Acentrallypositionedschematicdiagramofthehumanbodyshowstheentirenervoussystem.
Thesagitallysectionedheadfeaturesapartiallydissectedbrainandtransparentoverlaydefiningthepartsand
functions.Aroundtheouteredgeoftheplaque,twogreatlyenlargednervecellsshowthedendrites,axon,and
synapse,helpingstudentsunderstandthenervoussystem’sresponsetostimuli;touch isshownasanexample.
Itcomeswithanactivitybindercontaining lessonplans,copymastersforstudentactivities,andanoverhead
transparency.Size:24"Wx18"H.
$154.00
I...,372,373,374,375,376,377,378,379,380,381 383,384,385,386,387,388,389,390,391,392,...1012
Powered by FlippingBook