Ward's Biology 2016-2017 - page 155
LivingMaterials
|
Protists
155
•Classof35Students:
Testtubeortwo
ounce jar.
AllculturescomewithaLivingMaterialsCD-ROM inludingaPDFofworking
withAlgaeandProtozoaandcontainscomprehensivebackground information
withcareandhandling instructions.
Ward’s
Packaging
Options
Note:
Manyalgaeculturesare
culturedandshippedintesttubes
toensurepurityandquality.
Ward’sGuarantees/Shipping
•Ward’sUnconditionalSatisfactionGuarantee:
Ifyou’renotsatisfiedwith
theconditionofyourculture,we’llreplace itorgiveyouafullrefund.
• Mostcultures,exceptthosenoted,willbedeliveredtoyou intwobusiness
days.
Ward’sProtistCultures
•Ward’sprovidesagenerousamountofculturetoensureyouhaveenoughto
meettheneedsofyourentireclass.Ourclass-sizedculturesare in2oz.contain-
ers;manyothersuppliersprovidecultures in1oz.containers.Ward’s100-student
culturesize isequivalenttofourclass-sizedculturesofothersuppliers,andcosts
upto40% less.
• Allculturesaremaintained inourown labs.Thismeansthetechniciansthat
workdirectlywiththeculturesareavailabletoanswer yourquestions.
•Ward’scancontrolhowandwhenyourordergetsshippedtobestmeetyour
needsandschedule.
BrownAlgae
Kelpandseaweed;withadistinctsetofphotosyntheticpigments.
Dictyota
$8.19
Marine.Macroalga.Ribbon-likeblades.Collected.
Ectocarpus
$8.19
Marine.Filamentous.Branchedthallus,zoospores.Erdschreiber’smedium.
Fucus
$32.19
Marine.Macroalga.Bladderwrack.
Laminaria
$30.99
Marine.Macroalga.Kelp.Largespecimen.Collected.
Diatoms
Characterizedbyshells(tests)madeofsilica, intwoparts.
Cyclotella
$8.19
$15.49
Marine.Unicellular;small,centriccells.Marinemedium.
Culturefor
35Students
5ReadySlides
Culturefor
35Students
Thalassiosira
$8.19
Marine.Unicellular;brackish,centriccells.Marinemedium.
Blue-GreenAlgae
SEEBACTERIASECTIONFORCULTURELIST
Navicula
$8.19
Freshwater.Unicellular;pennate,withbilateralsymmetry.Basicculturesolution.
Synedra
$8.19
Freshwater,Unicellular;pennate,withbilateralsymmetry.Diatomorbasicmedium.
•5ReadySlides:
Cultureyour
protists ina7mLreusable
poly-propylenecontainerwitha
taperedend.ReadySlidesofferthe
convenienceofviewingcultures
underamicroscopewithoutthe
needtoprepareawetmount.
•Classof100Students:
Threetesttubesorone
eightounce jar.
ProtistCultures
“Ihavebeenaneducatorfor22years.Ofthat,
Ihavetaught/usedliveprotistsfor14years.
ThelasttwoyearsItried,tonoavail, two
differentvendorsinsearchofliveprotistcul-
turesthatwereconcentratedenoughformy
middleschoolstudentstoeasilylocateand
drawthem.Thisyear,ItriedWard’scultures.
Theywere,byfar,thebestculturesIhave
everused.Theywerealiveconcentrated,
andpure(someoftheculturesIusedlast
yearhadtwoorthreeprotistsotherthanthe
onesthatweresupposedtobeinthere).I
amsopleasedthat,asadepartmentchair,
Iamrecommendingyourculturestoallthe
departmentchairsinmycounty.Thankyou
forprovidingaqualityproduct.”
~
TracieMoots7thGradeLifeScience,
ScienceDepartmentChair
ShareyourreviewforyourfavoriteWard’s
Scienceproductsat
wardsci.com
Look for the reviews tabon everyproduct page.
I...,145,146,147,148,149,150,151,152,153,154 156,157,158,159,160,161,162,163,164,165,...1012
Powered by FlippingBook