Ward's Biology 2016-2017 - page 154
MicrobiologicalMedia
|
LivingMaterials
154
SlantRack
Theuniqueendplateon
thisrackallowsyouto
createagarslanttubesat
either5°or20°depending
onwhichsideyouuse.To
createtheslanttubes,sim-
plyautoclavethemedia
tubeswhiletheyare inthe
rack,andafterautoclaving,
settherackonthebenchatthedesiredangle.Themedia
willhardenwithaconsistentslant.Therack isalsousefulfor
incubating liquidculturesonaslant. Inaddition, it iskeyed
withnumbersand lettersfor identification, isstackable,and
doesnotfloat inwaterbaths. Itwillholdforty16mmtubes.
Size:11¼"x4½".
$61.44
Touch-FreeWaste
DisposalCan
ReducetheChanceofBiohazardCon-
tamination
Protectyourstudentsfromcontactwhenthey
aredisposingofbiohazardousmaterials.Thecan
automaticallyopensandcloseswhenactivatedby
asensor inthefront.Thebodyand lidaremolded
ofeasilycleanableABSplasticand inside isaremov-
able,autoclavable,poly-propylenepail. Itcanbe
usedwith14"x19"biohazardbagsandcomeswith
labelstodesignatethecanforbiohazard,trash,or
recyclable.Capacity:2.5gallon.
181410
DisposalCan
$95.00
NichromeInoculatingNeedle
andLoop
BothHeatQuicklytoSaveTime
Boththeneedleandthe looparemounted innon-breakable
aluminumhandles.Choosethefine-pointedneedleforsmall
coloniesorthe loopfor largerquantitiesofbroth.Size:each
10¾"L.
Needle
$3.95
Loop
$3.95
ColonyCounter
QuicklyEstimatethe
NumberofColonies
onaPlate
PlaceaPetridishoverthe
gridtoquicklyandaccurately
countcolonies.Thecounter is
Plexiglasandetch-printedwith
rulingsforusewith60or100mmdishes. Itcanbeusedwith
orwithoutanoutside lightsource.Size:5¼"Lx5¼"Wx4½"H.
$70.00
Anaerobic
GrowthBag
CreatesOxygen-Free
EnvironmentforGrowing
AnaerobicBacteria
Getthebestyieldofanaerobic
bacteriabyculturingthem inthese
transparentbags.Eachholdsupto
six16x125mmtubes.Thepackage
offivecomeswithagasgenerator,
catalyst, indicator,and instructions.
$45.99
SterileDisposablePetriDishes
Polystyrenedisheswithcovers.
Dia.xDepth Quantity Price
BulkQty.
BulkPrice
60x15mm 20
$7.32
500
$176.41
100x15mm 20
$10.58
500
$254.08
100x20mm 20
$17.85
400
$348.00
150x15mm 20
$38.84
120
$182.85
MicrobiologyMaterials
CultureTubeswithScrewCaps
Reusableglassculturetubescomewithscrewcapstopreventcontaminationor
leakageofbacteriologicalmedia.Autoclavable.Packageof100.
O.D.xLength Capacity
13x100mm 9mL
$2.25
16x150mm 20mL
$2.30
DisinfectingWipes
Sanitizeclassroomand laboratory
surfaceswiththesecleansingwipes
whichkill99.9%ofbacteria in30seconds.
Thirty-fivebleach-freewetwipes,
measuring7"x8"each,arestored ina
convenientdispenser.
151030
$8.20
PetriSticker™
CleverAlternative
toUsingColony
Counters
Simplyattachthistrans-
parent,waterproofsticker
tothebottomofyourPetri
dish.Theequally-shaped
divisionsofthegridare
idealforcolonycounts,
whilethe6-section is
perfectforsensitivitystudies.Packagesof36.
32-SquareGrid
$33.23
6SectorPie
$33.23
BiohazardBags
Instructionsareprintedoneverybiohazardwastedisposalbag,
to insureproperhandlingofhazardouswaste.Thesebagsare
polyethylene.Autoclavableto250°F.
BiohazardBags,12"x24",
Pkg.of100
$40.95
I...,144,145,146,147,148,149,150,151,152,153 155,156,157,158,159,160,161,162,163,164,...1012
Powered by FlippingBook