Ward's Biology 2016-2017 - page 153
LivingMaterials
|
MicrobiologicalMedia
153
PreparedMedia
Ward’sDehydratedMedia
DiluteQuantitiestoAnyVolumeasNeeded
Foraneconomicalalternative,useWARD’Sowndehydratedmediaformanymicrobiologyapplications.
Preparation instructionsare included.
Name
100gBottle Price 500gBottle Price
NutrientAgar
$42.20
$103.00
NutrientBroth
$44.10
$103.00
SabouraudDextroseAgar
$30.50
$76.75
TrypticSoyAgar
$31.50
$66.95
TrypticSoyBroth
$22.40
$50.50
DIFCO™DehydratedMedia
ConsistentlyHigh-QualityGrowthMedia
Thismedia iseasytomix.Justadddistilledwateraccordingtothe
included instructions.500gbottle.
BeefExtract
$136.00
BloodAgarBase
$175.54
Brain-Heart InfusionBroth
$154.50
EMBAgar(Levine)
$168.00
LitmusMilk
$170.19
MacConkeyAgar
$107.95
MannitolSaltAgar
$62.00
Mueller-HintonAgar
$108.50
NutrientAgar
$102.99
NutrientBroth
$103.50
PotatoDextroseAgar
$70.35
SabouraudDextroseAgar
$75.50
StarchAgar
$235.50
ThioglycollateBroth
$66.99
TrypticSoyAgar
$67.19
TrypticSoyBroth
$58.95
MediaBottles
Featuringa largeopening,no-drip lip,
andslopingshoulderstofacilitatepour-
ingandcleaning,thesebottlesshow
approximatevolumegraduationsand
havea largewritingarea.Boththebottles
andrubber-linedcapsareautoclavable.
Madeofborosilicateglass.
125mL
$7.55
250mL
$9.00
500mL
$12.60
MicrobiologyMaterials
Disposable
Bacti-Spreaders
IdealforBeginningMicrobiologyStudents
It’saneconomicalalternativetoatraditionalmetal
Bacti-Spreader;thisplastic instrument’s“L”shape
makes iteasiertodistributebacteriaovertheentire
plate.Size:5¾"Lx1½"W.Packageoften.
$4.40
Bacti-Spreader
SpecialDesignEvenlyDistributesBacteria
Thestainlesssteelrodhasbeenspecificallydesigned
forgreatercontactwithagarformoreevendistribu-
tionofbacteria.Size:9"Lx2"W.
$5.00
SwabApplicators
ForEasySubculturingand
BacterialTransfers
Sterile,cotton-tippedapplicatorsspreadaconfluent
lawnofbacteriawithoutbreakingtheagarsurface.
Individuallywrapped.Size:6"L.
Pkg.of10
$4.95
Pkg.of100
$17.50
DisposableInoculating
Loop/Needle
TwoToolsinOne
These individuallywrapped,sterileplastictoolsdon’t
needheating.Eachfeaturesa loopatoneendanda
needletipattheother.Size:8"L.Packageof25.
$10.00
I...,143,144,145,146,147,148,149,150,151,152 154,155,156,157,158,159,160,161,162,163,...1012
Powered by FlippingBook