Ward's Biology 2016-2017 - page 152
MicrobiologicalMedia
|
LivingMaterials
152
PreparedMedia
TotalCount (SPC)Easygel
PotatoDextroseAgar
NutrientAgar
Easygel™Media
ConvenientOne-StepProcedure
Useful inawiderangeofapplications,Easygel isasterile,premixed,premeasuredgrowthmediumthatrequires
nomixing,noautoclaving,andnowaterbaths.SimplypourEasygel intothe includedpretreatedPetridish;
itgelsquicklyand isvirtually identical inappearancetoagarmediaplates.Easygelcanbestoredforoneyear
refrigerated.EachMediaPackcomeswithtenor40pretreatedplates,enoughEasygeltopourthetotalnumber
of includedplates,and instructions.
ColiscanEasygel
$43.20
$117.35
Fordifferential isolationofGram-negativebacteria.
10Plate Price
30Plate
40Plate
Name
MediaPack Price MediaPack Price MediaPack Price
NutrientEasygel
$70.20
$70.20
Multipurposemedia.Supportsthegrowthofavarietyofmicroorganisms.
TotalCount(SPC)Easygel
$69.20
Standardplatecountmedia.Forenumerationofcoliformbacteria,etc.
TrypticSoyEasygel
$25.49
$81.45
Multipurposemedia.Supportsgrowthofnumerousbacterialspecies.
DehydratedMedia
PremeasuredPacksSaveTimeintheLab
SimplyaddthecontentsofoneDehydratedMediapacktoone literofwater,bringtoaboil,andsterilize,to
makeenoughmediafor40to50platesor160tubes.
BileEsculinAgar
Fordifferential isolationandpresumptive
identificationofgroupDstreptococcus.
$19.15
Mueller-HintonAgar
Fortestingantimicrobialsusceptibility
ofmicroorganismsbyagar
diffusionmethod.
$19.15
NutrientBroth
Multipurposemedia.Supportsthegrowth
ofavarietyofmicroorganisms.
$19.15
PotatoDextroseAgar
Forcultivationoffungiandyeasts.
$19.15
SabouraudDextroseAgar
Forcultivationofyeasts,fungi,andaciduric
organisms.
$15.00
TrypticSoyAgar
Soybean-caseindigestmedium.Supports
thegrowthofavariety
ofmicroorganisms.
$19.15
TrypticSoyBroth
Soybean-caseindigestmedium.Supports
thegrowthofavariety
ofmicroorganisms.
$20.19
NutrientAgar
Multipurposemedia.Supportsthegrowth
ofavarietyofmicroorganisms.
$19.15
MotilityTestAgar
Showsgliding,flagellate,andcorkscrew
motionofbacteria.
$26.00
“Thisisthebestandeasiesttouseagar.Unlikemostpre-pouredagaritdoesnothavetobe
refrigerated.Icanstoreitalongtime.Iuseiteveryyearwithmy5thgraders.Itgrowsbacteria
andfungieasily.Allyouhavetodoispourthepremeasuredliquidintothespeciallytreated
petridishesthatareinthebox,wait45minutesandit'sreadytouse!”
~Ms.D,DuchesneSchoolDistrict
ShareyourreviewforyourfavoriteWard’sScienceproductsat
wardsci.com
Look for the reviews tabon everyproduct page.
I...,142,143,144,145,146,147,148,149,150,151 153,154,155,156,157,158,159,160,161,162,...1012
Powered by FlippingBook