Ward's Biology 2016-2017 - page 151
LivingMaterials
|
MicrobiologicalMedia
151
PreparedMedia
Tube
Twelve16x125mmscrewcaptubes.Slanted
unlessnoted.
MediaPourPak
Packageof20sterilePetriplates(90x15mm)
andthreemediabottles.Eachbottlepours
sevenplates.
Plates
Ten90x15mmplates,sealed.Canbestoredtwoweeks.
Bottle
Six125mmbottles.Suf-
ficientfor15–20tubesor
5-7platesperbottle.
BloodAgar
Forcultivationoffastidiousorganismsand identificationof
hemolyticreactions.
Note:
Soldonlytocolleges.
Pkg.of12Tubes
$27.19
Pkg.of10Plates
$23.50
LitmusMilkBroth
Fordemonstrationofacidproduction,coagulation,
orproteolysisofcasein inculture identification.
Pkg.of12Tubes
$23.99
Lowenstein-JensenAgar
For isolationand identificationofMycobacteriumspecies.
Pkg.of12Tubes
$35.99
MannitolSaltAgar
Forcultivationofpathogenicstaphylococci.
Pkg.of12Tubes
$21.19
ChocolateAgar
SupplementedwithXandVfactors.Forcultivationof
Neisseria
and
Haemophilus
species.
Pkg.of12Tubes
$26.99
HalobacteriumAgar
Forcultivationofhalophilicbacilli.
Pkg.of12Tubes
$44.99
LuriaBroth
Completenutrientbrothforgrowing
E.coli
cultures.
Pkg.of12Tubes
$23.49
Pkg.of6Bottles
$25.99
Blood,Bovine
Defibrinated.Use inbloodcomparisonstudiesorblood
agar.
Note:
Soldonlytocollegesandaccredited labs.
100mLBottle
$53.10
Blood,Sheep
Defibrinated.UsewithBloodAgarBasetomakeBlood
Agar.100mLbottle.
Note:
Soldonlytocollegesand
accredited labs.
100mLBottle
$56.25
ColumbiaCNAAgar
With5%sheep’sblood.For isolationofGram-positive
organisms.
Note:
Soldonlytocollegesandaccredited labs.
Requires24hournotice.
Pkg.of10Plates
$25.49
Agar,PlainPowder
25gBottle
$8.95
100gBottle
$19.50
500gBottle
$64.79
LuriaAgar
Completenutrientagarforgrowing
E.coli
cultures.Eachof
thebottles inthepackageofsix is200mL. Includesempty
plates.
Pkg.of6Bottles
$44.99
Pkg.of10Plates
$19.90
MacConkeyAgar
Fordifferential isolationofentericbacillibasedon lactose
fermentation.
Pkg.of10Plates
$20.19
Brain-HeartInfusionAgar
Forcultivationoffastidousorganisms;e.g.,streptococci,
pneumococci,andmeningococci.
Pkg.of10
$27.99
EMBAgar
Fordifferential isolationofGram-negativeorganisms.
Pkg.of6Bottles
$40.00
Pkg.of10
$24.49
NutrientAgar
General-purposemedia.Supportsthegrowthofa
varietyofmicroorganisms.
Pkg.of12Tubes
$18.99
Pkg.of6Bottles
$43.19
Pkg.of10Plates
$21.50
PourPak
$22.99
NutrientAgarDeeps
Useforasinglepourplate.15mLpertube.
Pkg.of12Tubes
$20.49
NutrientBroth
General-purposemedia.Supportsthegrowthofavariety
ofmicroorganisms.
Pkg.of12Tubes
$18.49
Pkg.of6Bottles
$39.19
PhenolphthaleinAgar
Forosmosisanddiffusionexperiments.
800mLBottle
$33.99
Non-NutrientAgar
Nonutrientsadded.Forcontrolplates.
Pkg.of10Plates
$24.49
TrypticSoyAgar
Soybean-caseindigestmedium.Supportsthegrowthofa
varietyofmicroorganisms.
Pkg.of12Tubes
$19.99
Pkg.of6Bottles
$46.49
Pkg.of10Plates
$21.95
PourPak
$20.99
SkimMilkAgar
Fordifferentiationoforganismsbasedonproteolysisof
caseinandcoagulation.
Pkg.of10Plates
$24.49
Sordaria
Agar
Forgrowthandsporulationofascomycetes,specifically
Sordaria
species.
Pkg.of6Bottles
$37.19
Sordaria
MatingAgar
Supportsandenhancesmatingof
Sordarias
trainsfor
geneticstudies.
Pkg.of6Bottles
$37.19
Pkg.of10Plates
$30.50
StarchAgar
Forstarchhydrolysistests
Pkg.of6Bottles
$27.49
Pkg.of10Plates
$26.99
ThioglycollateMedium
Without indicator.Forcultivationofanaerobic
speciesand
Clostridium.
Suitableforfermentation
studieswhereno indicator isrequired.
Pkg.of12Tubes
$25.19
TomatoJuiceAgar
Forcultivationof lactobacilliandother
acidophillicorganisms.
Pkg.of12Tubes
$20.99
TrypticSoyAgar,Soft
Foroverlays inplaquemorphologystudies.
Pkg.of12Tubes
$19.49
TrypticSoyBroth
Soybean-caseindigestmedium.Supportsthegrowthofa
varietyofmicroorganisms.
Pkg.of12Tubes
$22.99
Pkg.of6Bottles
$38.49
TSIAgar
For identificationofGram-negativebacteriabasedon
dextrose, lactose,andsucrosefermentation,andhydrogen
sulfideproduction.
Pkg.of12Tubes
$17.99
Yeast,Genetic(YEADAgar)
Yeastextract-adenine-dextrosegrowthmedia.Formain-
taininghaploidcultures.
Pkg.of6Bottles
$52.49
Yeast,Genetic(YEDAgar)
Yeastextract-dextrosegeneralgrowthmedium.For
demonstrationofredorwhitepigmenttraits.
888902
Pkg.of6Bottles
$28.49
Pkg.of10Plates
$22.49
SabouraudDextroseAgar
Forcultivationofyeasts,molds,andaciduricorganisms.
Pkg.of10Plates
$22.95
SabouraudDextroseBroth
Forcultivationofyeasts,molds,andaciduricorganisms.
Pkg.of12Tubes
$23.99
SIMMedium
ForthedifferentiationofmembersoftheEnterobacteriacaea
basedonH
2
Sproduction, indoleproduction,andmotility.
Pkg.of12Tubes
$20.49
YMAgar
Yeastmaltagarmaintenancemediafor
Schizophyllum
species.
Supportsfungalgrowth.
Pkg.of4AgarJars
$27.30
PotatoDextroseAgar
Forcultivationofyeastsandmolds.
Pkg.of6Bottles
$31.99
Pkg.of10Plates
$24.75
Ward’scancustompreparemedia
thatyoudon’tsee listed.Formore
information,call
800-962-2660
.
I...,141,142,143,144,145,146,147,148,149,150 152,153,154,155,156,157,158,159,160,161,...1012
Powered by FlippingBook