Ward's Biology 2016-2017 - page 150
MicrobiologicalMedia
|
LivingMaterials
150
Ward’sBreadMoldCultureandStudyKit
CultureandIdentifyDifferentTypesofMold
Beginningwiththe initialculture,grownonslicesofbread,studentsobservethe
developmentofmold,thenusethe includeddichotomouskeyto identifythetypes
ofmoldthatappear,aswellascomparetheirgrowthratesandmorphological
characteristics.Theactivitycanbecompletedwithoutcookingmedia,dispensing,or
steriletechnique.Thekit includesdemonstrationplateculturesof
Aspergillusniger,
Mucorheimalis,Penicilliumnotatum,
and
Rhizopusstolonifer;
culturematerialsfor15
labgroups;teacher’sguide;andstudentcopymasterwithdichotomouskeyand
additionalsuggestedactivities.
Note:
Includesperishablematerials.Pleasespecify
deliverydate.
$88.50
Ward’sPilobolusCultureandStudyKit
ObserveaDramaticExampleofPhototropism
Thisfungushastheabilitytoshoot itssporangiatowarda lightsource,
accuratetowithinafewmillimeters,wheretheyadheretoanysolidob-
jectstheystrike.Thiskitprovidesallthenecessarymaterialsforstudents
toobservethis illustrationofpositivephototropismright inthe lab.The
kitcomeswithatubeculture of
Piloboluscrystallinus
, instructions,and
materialsforthree labgroups.
Note:
Includesperishablematerials.Please
specifydeliverydate.
$41.49
Ward’sPredatoryFungiKit
WatchaFungusTrapandDigestaRoundworm
Foradramaticdemonstrationofpredatorybehavior,addanematode
tothecultureoffungusyougrow.Thefungus,commonlyfound insoil,
hasadhesive loopsattheendsof itshyphalbranches.Asthenematode
passesthrougha loop, itbecomestrapped,andthefungusdigests it. It
includesculturesof
Arthrobotrysconoides
and
Rhabditis,
instructions,and
materialsforaclassof30students.
Note:
Includesperishablematerials.
Pleasespecifydeliverydate.
$48.19
GrowingGourmetMushroomsLabActivity
GrowExotic-LookingMushrooms
Presentacomprehensiveunitonfungibygrowingoystermushroomsandwatching
themtransformsawdust intonutritioussoil.Yourstudentswill learnthe importance
offungi,howtheyreproduce,andthestructureandfunctionofalltheparts. In
addition,they’ll investigatetheeffectsofgravity(geotropism)onmushroomgrowth
adaptiveresponsesandthe implicationforfungalreproduction. Thekitcontains
onecontrolandonetreatmentsetup,and literaturewithcorrelationtothenational
sciencestandards.
Note:
Includesperishablematerials.Pleasespecifydeliverydate.
OnlyshippedonMondaysandTuesdays.
$41.85
FungiDiversity
Can’tfindwhatyou’re lookingfor?
Wehaveanevengreaterselectionofsuppliesfor
everysciencesubjectandgradelevelonline.
Seeit
allonline!
I...,140,141,142,143,144,145,146,147,148,149 151,152,153,154,155,156,157,158,159,160,...1012
Powered by FlippingBook