Ward's Biology 2016-2017 - page 149
LivingMaterials
|
Fungi
149
Lichens
FolioseLichen
FruiticoseLichen
CrustoseLichen
Ward’sLichenTypes
Oneeachoffoliose,crustose,and
fruticose lichen.Thefolioseandcrustose
lichensareanyofawidevarietyofspe-
cies.Allsamplesarefield-collectedand
come ina4oz. labeled jar.Setofthree.
$18.49
Ward’sFungi
Classification
Set
Representatives
fromtheFourMajor
FungalGroups
Sixcultures,allgrown in
ourown labsandsealed
attheoptimalpointof
fungalgrowth,clearly
illustratethestructural
differencesbetweenthe
fourfungalgroups.Dem-
onstrationplatecultures:
Schizophyllumcommune
(Basidiomycete);
Aspergillus
niger,Penicilliumnotatum
(Deuteromycete);
Mucorheimalis,Rhizopusstolonifer
(Zygomycete);and
Neurosporacrassa(
Ascomycete).Thesetofsixcomeswith
informationonfungalstructureandsuggestedactivities,aswellasthe
Working
withFungi
manual.
$49.99
Ward’sFungi
IdentificationSet
IdentifySpecimens
withIncluded
DichotomousKey
Theserepresentativesofthe
fourmajorfungalgroups,plusa
specimenof lichenforadditional
study,allowstudentstocompare
theirvariousstructuresand
classifyeachtype.The included
dichotomouskeyeasily leads
studentsthroughthe identificationprocess.Fungispecimensaresuppliedas
demonstrationplates;the lichen issupplied ina jar.Cultures:
Schizophyllum
commune
(Basidiomycete),
Penicilliumnotatum
(Deuteromucete),
Rhizopus
stolonifer
(Zygomycete),
Saccharomycescerevisiae
(Ascomycete),andFruticose
lichen.Thesetoffive includes informationonfungalstructureandsuggested
activities,aswellasthe
WorkingwithFungi
manual.
$59.99
Ward’s
HighSchool
FungiSet
ExamplesofMost
CommonTypes
FeaturedinHigh
SchoolTextbooks
Enhanceyour lessons
withreal samplesof the
fungidepicted inmost
highschoolbiology
texts; thesespecimens
makeupacomprehen-
sivesurvey ideal for
introductorystudiesof fungi.The12 fungi specimensaresupplied
astubeculturesanddemonstrationplates.Cultures:
Arthrobotrys
conoides,Aspergillusniger,Mucorrouxii,Neurosporacrassa,Pencillium
notatum,Phycomycesblakesleeanus,Rhizopusstolonifer,Rodotorula
rubrum,Saccharomycescerevisiae,Schizophyllumcommune,Sordaria
fimicola,
and
Trichodermaviride.
$119.19
FungiSets
Ward’s
Zygospore
FormationSets
ObserveResultsofFungal
SexualReproduction
Yourchoiceoftwosexually
reproducingfungiareavailable
indemonstrationplates;eachset
containsa“+”strainononehalfof
theplateanda“-"strainontheother
half,witha lineofzygosporesalready
formedatthe junctionofthemycelia.Each
setcomeswithateacher’sguideandthe
Working
withFungi
manual.
Mucorheimalis
(+)and(-)
$25.19
Phycomycesblakesleeanus
(+)and(-)
$25.19
Ward’s
Sordaria
Genetics
LabActivity
ClassicExperimentto
DemonstrateCrossingOver
Studentswillbeableto identify
geneticdifferencesbetweendiploid
andhaploidorganisms,explainhow
geneticvariabilityarisesfromsexual
reproduction,andstudytheroles
ofmitosisandmeiosis.Studentswillgrowwildtypeandmutant
Sordaria
strainsonmatingagarandexaminetheascosporesunderamicroscope
(availableseparately).
$90.75
Formoregeneticsactivitiesinvolving
Sordariaandyeast,looktotheGenetics
sectionofthiscatalogorvisitusonline.
I...,139,140,141,142,143,144,145,146,147,148 150,151,152,153,154,155,156,157,158,159,...1012
Powered by FlippingBook