Ward's Biology 2016-2017 - page 148
Fungi
|
LivingMaterials
148
FungiCultures(cont.)
Name
TubeCulture
DemoPlate
$12.19
$13.49
Saccharomycescerevisiae
var.
ellipsoides
Ascomycete.Wineyeast:Used infermentationactivities. Incubationtemperature30°C.Sabourauddextroseagargrowthmedium.
Rhodotorularubra
Rhizopusstolonifer,nigricans
Schizophyllumcommune
FungiCharts
PhotosandPhotomicrographsShowFungiStructures
Vividcolormicrophotographsshowcharacteristicsoffungi indetailedrepresentation. Examinestructures
ofvarioustypesoffungiordiscernthedifferentstepsofbothsexualandasexualphases.Eachposteroffers
studentsaclose-upviewoffungiandhelps leadtodeeperunderstanding.Laminated.Size:18"x24".
AscomycetesChart
$27.50
BasidiomycetesChart
$27.50
ZygomycetesChart
$27.50
Sordariafimicola
(GrayMutant)
Ascomycete.Crosseswithwildortanstrains;recommendedforgeneticstudies. Incubationtemperature25°C.
Sordaria
agargrowthmedium
Sordariafimicola
(TanMutant)
Ascomycete.Crosseswithwildorgraystrains;recommendedforgeneticstudies. Incubationtemperature25°C.
Sordaria
agargrowthmedium
Sordariafimicola
(WildType)
Ascomycete.Crosseswithtanorgraystrains;recommendedforgeneticstudies. Incubationtemperature25°C.
Sordaria
agargrowthmedium
Saccharomycesa1,
WildType(HAO)
Incubationtemperature30°C.YEADagargrowthmedium.
Saccharomyces
a
1,
WildType(HBO)
Incubationtemperature30°C.YEADagargrowthmedium.
Saccharomycesa2,
Red(HAR)
Adenine-dependent.RedpigmentonYEDagargrowthmedium.
Saccharomyces
a
2,Red(HBR)
Adenine-dependent.RedpigmentonYEDagargrowthmedium.
Saccharomycesa3
(HAT)
Trytophandependent. Incubationtemperature30°C.YEADagargrowthmedium.
Saccharomyces
a
3(HBT)
Trytophandependent. Incubationtemperature30°C.YEADagargrowthmedium.
Saccharomycesa4,
Red(HART)
Trytophanandadeninedependent. Incubationtemperature30°C.YEADagargrowthmedium.
Saccharomyces
a
4
,Red(HBRT)
Trytophanandadeninedependent. Incubationtemperature30°C.YEADagargrowthmedium.
Forkitstostudy
yeastgenetics,see
theGeneticssec-
tionofthiscatalog.
Key:
Pathogen
n
PlantPathogen
Seethe introductionpageofFungiforanexplanationofpathogens.
I...,138,139,140,141,142,143,144,145,146,147 149,150,151,152,153,154,155,156,157,158,...1012
Powered by FlippingBook