Ward's Biology 2016-2017 - page 147
LivingMaterials
|
Fungi
147
Neurosporacrassa
Penicilliumnotatum
Aspergillusniger
FungiCultures
Name
TubeCulture
DemoPlate
$12.19
$13.49
Arthrobotrysconoides
Deuteromycete.Biologicalcontrolorganism.Trapsnematodes. Incubationtemperature25°C.Cornmealagargrowthmedium.
Aspergillusniger
Deuteromycete.“Blackmold".Producescitricacid.Commonairbornecontaminant.Causesaspergillosis.Incubationtemperature25°C.Sabourauddextroseagargrowthmedium.
• 
Candidaalbicans
Deuteromycete.Causesthrush,candidiasis.Multipliesbybudding.Formsgerm-tubes inserumcultures. Incubationtemperature37°C.Sabourauddextroseagargrowthmedium.
Coprinuscinereus
Basidiomycete. Inky-cappedmushroom.Jarculture. Incubationtemperature30°C.YM/rabbitdungagargrowthmedium.
Mucorhiemalis
(+)
Zygomycete.DungMold.Matingstrain;crosswith
Mucorheimalis(
-)forzygosporeformation. Incubationtemperature25°C.Potatodextroseagargrowthmedium.
Mucorhiemalis
(-)
Zygomycete.DungMold.Matingstrain;crosswith
Mucorheimalis
(+)forzygosporeformation. Incubationtemperature25°C.Potatodextroseagargrowthmedium.
Neurosporacrassa
Ascomycete.“Bakerymold";redbreadmold. Incubationtemperature25°C.Neurosporaagargrowthmedium.
Penicilliumchrysogenum
Deuteromycete.Producespenicillin. Incubationtemperature25°C.Maltextractagargrowthmedium.
Penicilliumnotatum
Deuteromycete.Bluemold;foodmold.Producespenicillin. Incubationtemperature25°C.Potatodextroseagargrowthmedium
Penicilliumnotatum
(High-YieldStrain)
Deuteromycete.Foodmold.Producespenicillin. Incubationtemperature25°C.Potatodextroseagargrowthmedium.
Piloboluscrystallinus
858671*
Zygomycete.“Shotgunfungus".Exhibitsphototropicresponse. Incubationtemperature25°C.*Dungagargrowthmedium, in jar.
Rhizopusstolonifer(nigricans)
(+)
Zygomycete.Commonblackbreadmold.Labcontaminant.Matingstrain,crosswith
Rhizopusstolonifer(nigricans)
(-)toproducezygospores. Incubationtemperature25°C.Potato
dextroseagargrowthmedium.
Rhizopusstolonifer(nigricans)
(-)
Zygomycete.Commonblackbreadmold.Labcontaminant.Matingstrain,crosswith
Rhizopusstolonifer(nigricans)
(+)toproducezygospores. Incubationtemperature25°C.Potato
dextroseagargrowthmedium.
Rhodotorularubra
Deuteromycete.Pinkyeast.Pinktoredpigmentonagargrowthmedium. Incubationtemperature30°C.Sabourauddextrose/YMagargrowthmedium.
Saccharomycescerevisiae
Ascomycete.HostforrDNA. Incubationtemperature30°C.Sabourauddextroseagargrowthmedium.
Saccharomycescerevisiae
Ascomycete.Baker’syeast.Reproductionbybudding. Incubationtemperature30°C.Sabourauddextroseagargrowthmedium.
I...,137,138,139,140,141,142,143,144,145,146 148,149,150,151,152,153,154,155,156,157,...1012
Powered by FlippingBook