Ward's Biology 2016-2017 - page 144
Monera
|
LivingMaterials
144
Ward’sSensitivity
TestDiscs
DetermineSusceptibilityorResistance
ofanOrganismtoSpecificAntibiotics
EasilyobservethesensitivityofGram-positiveorGram-
negativebacteriatosixdifferenttypesofantibioticswith
theseconvenientkits.Eachkitcontains50discsofeach
typeofantibiotic;allyouneedtodo isplacethemonthebacterial
cultureandobservetheresults. Instructionsare included.
A)GramPositiveDiscs
Contents:
chloramphenicol,erythromycin,novobiocin,penicillin,streptomycin,
andtetracyline.
$67.19
B)GramNegativeDiscs
Contents:
chloramphenicol,furadantin,kanamycin,nalidixicacid,tetracycline,
andtriplesulfa.
$67.19
AntibioticSensitivityDiscSet
Onevialcontains50discseachofpenicillin,streptomycin,
tetracycline,neomycin,anderythromycin.
$62.99
Sensi-Disc™Magazines
Eachmagazinecontains50discsofone
agent.Adisc dispenser,required, isavailable
separately.
Ampicillin,10
m
g
$14.95
Bacitracin,10units
$110.05
Chloramphenicol,30
m
g
$14.95
Erythromycin,15
m
g
$14.95
Kanamycin,30
m
g
$14.95
Neomycin,30
m
g
$14.95
Novobiocin,30
m
g
$14.95
Oxytetracycline,10units
$110.05
Penicillin,10units
$14.95
Streptomycin,10
m
g
$14.95
Tetracycline,30
m
g
$14.95
DispenserforSensi-Disc
Thisstainlesssteeldispenserreleasesoneantibioticdiscata
timefromaSensi-DiscMagazine.
$34.00
AntibioticSensitivity
Blank
SterileDiscs
Sterile,6mm-diametersterile
discsareexcellentforuseas
controldiscs.Bottleof100.
$13.99
A
B
Microbiology
FlashCards
EncouragesRetentionWhile
EnhancingRapidReview
Thesedurablecardsareperfectmedical
referencesforhighereducation.These
cardsareuseful indistinguishingthe
differencesbetweenviruses,bacteria,
protozoa,andfungi,aswellascreating
anunderstandingaboutthecauseof
thediseasesandthemethodstocontrol
them.Labeledstructuresarepresented
onthefrontandconceptsareonthe
back.Each is4¼"x5½".Setof150.
$34.95
EndosporeStainKit
Distinguishbetweenendosporesandthevegeta-
tivestructuresofbacterialcells.Theendospores
willstaingreenagainstapinkbackground. In-
cludessporestainingsolution,safraninOsolution,
and instructions.
Stains50Slides
$11.90
Stains750Slides
$22.50
I...,134,135,136,137,138,139,140,141,142,143 145,146,147,148,149,150,151,152,153,154,...1012
Powered by FlippingBook