Ward's Biology 2016-2017 - page 141
LivingMaterials
|
Monera
141
StaphyloslideLatex
TestKit
FirstStepinIdentifyingPathogenicStaph
Apositiveagglutinationreaction isaneasywaytodetectpathogenic
Staphy-
lococcus.
Thiskitgivesyouresultswithinfiveminutesandcomeswithenough
materialsfor100tests. It includestwo2.5mLbottleseachoftestandcontrol
latexand35reactioncards.
Note:
Reagentscontainmaterialofhumanorigin.
$164.30
IdentifyingBacteria
Ward’sNGSSActivityKit:
BiochemicalScreeningofBacteria
IdentifyUnknownBacteriaUsingThreeActualLaboratoryTests
Studentswillperformagrossexamination,Gramstaining,andstudyunderthemicroscope,as
wellasatestof thebacteria’schemicalproperties—eachtest,employed inrealmicrobiology
labs,helpsto identifyunknownbacteria.ThechemicaltestusestheEnterotube IIsystem,anaccu-
ratetestmethodthatperforms12standardizedtest inonestep.Demonstrationplates:Enterobacter
aerogenes,E.cloacae,Escherichiacoli,Serratia liquefaciens,andS.marcescens. Itcomeswithenough
materialsforfive labgroups,ateacher’sguide,andstudentcopymaster.
Note:
Includescouponfor
perishablematerials.Pleasespecifydeliverydate.Allowtwoweeksfordelivery.
$199.75
Ward’sAcid-FastStainingLabActivity
IdentifyandDifferentiateBacteria
Acid-faststain isusedprimarily inthe identificationofpathogenicorganisms
likethetuberculosisbacillus,
Mycobacteriumtuberculosis
,andthe leprosy
microbe,
Mycobacteriumleprae.
Inthisactivity,studentsperformanacid-fast
stainprocedureonnonpathogenicculturesof
Mycobacteriumnonchromo-
genicum
,anacid-fastrodfound insoil,and
Staphylococcusepidermidis
,a
non-acid-fastcoccusthatnormally inhabitsyourskin. Intheprocess,students
will learntousesteriletechniques,oneofthemostcrucialcomponentsof
microbiology.Theywillalsorelatethecharacteristicsofacid-fastbacteria
todrugresistance. It includesateacher’sguide,studentcopymaster,and
materialsforeightsetups.Acompoundmicroscopewithanoil immersion
lens,required, isavailableseparately.
Note:
Coupon includedforperishable
materials.Redeembymail,fax,phone,ore-mail.
$107.10
DrySlides™
Fast,ConvenientMethodfor
BiochemicalTestingofBacteria
Oxidase isanacidometricprocedureforthedetectionofbeta-lactimaseproduc-
tion.Eachslidecontainsfourreactionareasforperformingmultipletests.Simply
smeara littleoftheunknowncolonyontothecoated35mmslidesandwithin
30secondsto2minutes,dependingonthereaction,youwillseeresults.A
positiveresult is indicatedbyacolorchange.Comeswith instructions.Package
ofnine.
$37.55
+
UsetomeetNGSSLS1:
FromMoleculestoOrganisms:StructuresandProcesses
DiscoverMoreProductsOnline!
We’vegotevenmoreproductsandsolutionstohelp
youturnsciencelessonsintoscienceconnections
online.
Shopthefullselectiononline.
I...,131,132,133,134,135,136,137,138,139,140 142,143,144,145,146,147,148,149,150,151,...1012
Powered by FlippingBook