Ward's Biology 2016-2017 - page 138
Monera
|
LivingMaterials
138
BacteriaSets
Ward’sHighSchool
BacteriaSet
AlltheMostCommonBacteriaFeatured
inSecondarySchoolTextbooks
Thissetofculturescanbestudiedashands-on
examplesofthebacteriadescribed inthemost
popularhighschool-levelbiologytexts.Tubecultures:
Aquaspirilliumserpens,Bacillusmegaterium,Clostridium
sporogenes,Escherichiacoli,Halobacteriumsalinarium,
Kocuriarhizophila,Lactobacilluscasei,Lactococcuslactis,
Pseudomonasfluorescens,Rhodospirillumrubrum,
Serratiamarcescens
D1, and
Vibriofischeri
.Thesetof12
comeswiththe
WorkingwithBacteria
manual.
$120.19
Ward’sBacteria
FormsMixture
ThreeMorphologyTypesinOneTube
Studentscannowcompareallthreemorphology
typeswithoutcreatingseparateslides.Onetube
contains:
Bacillussubtilis
(rod),
Kocuriarhizophila
(cocci),and
Aquaspirillumserpens(
spirillum).Sup-
plied intrypticsoybroth.Theset includesan
Explano-Mount,which isamicroscopeslidewith
an illustratedguidethathighlightsallthe important
informationontheslide,andthe
Workingwith
Bacteria
manual.
$20.19
Ward’sBacteriaFormsSet
IncludesExamplesoftheThreeBasic
MorphologicalTypes
Viewallthreetypesofbacteria individually,then
comparetheirdifferentstructures,withtheselection
ofspecimens inthisset.Tubecultures:
Bacillussubtilis
(rod),
Rhodospirillumrubrum
(spirillum),and
Kocuria
rhizophila
(coccus).Thesetofthreecomeswiththe
WorkingwithBacteria
manual.
$34.99
Ward’sOil-Degrading
MicrobesSet
StudytheMicrobesUsedInthe
BioremediationofOilSpills
Thisuniquesetfeaturesnaturallyoccurring
microbesthathaveanaffinityfordegradingpetro-
leumproducts,makingthemuseful intheprocess
ofcleaningupoilspills.Theculturesaresupplied
freeze-dried,withmedia.Cultures:
Penicillium
sp.
and
Pseudomonas
sp.Thesetoftwocomeswiththe
WorkingwithBacteria
manual.
$44.49
Pathogen
Seethe introductionpageofMonera foranexplanationofpathogens.
Ward’sCollegeBacteriaSet
SpecimensPresentedMostOften
inUniversityTextbooks
Acomprehensivesetofavarietyofbacteria,
theseculturesarean ideal supplementtotext-
bookmicrobiologystudies.Freeze-driedcultures:
B.subtilis,Bacillusmegaterium,C.rubrum,C.
pseudodiphtheriticum,C.xerosis,Clostridiumsporo-
genes,Corynebacteriumdiphtheriae,Edwardsiella
tarda,Enterobacteraerogenes,Enterococcusfaeca-
lis,Escherichiacoli,Klebsiellapneumoniae,Lactoba-
cilluscasei, Lactococcus lactis,Morexellacatarrha-
lis,Mycobacteriumphlei,M.smegmatis,Neisseria
sicca, P.hauseri,Proteusmirabilis,Pseudomonas
aeruginosa,Salmonellaenteritidis,S.epidermidis,S.
pneumoniae,S.pyogenes,S. typhimerium,Serratia
marcescens,Shigellaflexneri,andStaphylococcus
aureus
(coagulasepositive).Thesetof29comes
withthe
WorkingwithBacteria
manual.
$429.99
Ward’sCyanobacteria
MorphologySet
StudyStructuralVarietyin
Blue-GreenAlgae
Thiscollectionofdifferenttypesofcyanobacteria
displaystheirstructuraldiversity.Cultures:
Oscil-
latoria,Nostoc
(filamentous);
Gloeocapsa
(colonial
globular);and
Gloeotrichia
(colonialfilamentous).
Thesetoffour includesthe
WorkingwithAlgae
manualandfourpipets.
$30.19
Ward’sCyanobacteria
Mixture
BroadSampleofSeveral
TypesofBlue-GreenAlgae
Fiveormoreofthemorecommontypesofcyano-
bacteriaaresupplied;specimensvary.Themixture
comeswiththe
WorkingwithAlgae
manualand is
supplied inasingle jar.
Culturefor35Students
$14.99
Culturefor100Students
$37.19
CyanobacteriaSets
I...,128,129,130,131,132,133,134,135,136,137 139,140,141,142,143,144,145,146,147,148,...1012
Powered by FlippingBook