Ward's Biology 2016-2017 - page 137
LivingMaterials
|
Monera
137
EubacteriaCultures(cont.)
Freeze-DriedCulture
withTransfer
Name
TubeCulture
andGrowthMedia
$11.19
$19.19
Streptococcusequisimilis
LancefieldgroupC,betahemolysis.Causesstrangles inhorses.Maybepathogenic inhumans.Camptestnegative;resistantto
bacitracin,doesnotferment lactose.Nonmotile.Cocci;pairsandchains.Grampositivereaction.Aerobic. Incubationtemperature
37°C.Bloodagargrowthmedium.
Streptococcusmutans
Foundondentalplaque, implicated incavities.NoLancefieldgroupshown,gammahemolysis.Nonmotile.Cocci;chains.Gram
positivereaction.Aerobic. Incubationtemperature37°C.Trypticsoyagar/bloodagargrowthmedium.
Streptococcuspneumoniae
NoLancefieldgroupshown,alphahemolysis.Bilesolubilitytestpositive,optochinsensitive.Causespneumonia.Nonmotile.
Cocci;chains.Grampositivereaction.Aerobic. Incubationtemperature37°C.Bloodagargrowthmedium.
Streptococcuspyogenes
LancefieldgroupA,betahemolysis.Sensitivetobacitracin.Causesstrepthroat,rheumaticfever,andscarletfever.Nonmotile.
Cocci;chains.Grampositivereaction.Aerobic. Incubationtemperature37°C; italsogrowswell in5%CO
2
.Bloodagargrowth
medium.
Streptococcussalivarius
Normalfloraofthemouthandthroat.LancefieldgroupK,gammahemolysis.Nonmotile.Cocci;chains.Grampositivereaction.
Aerobic. Incubationtemperature37°C.Bloodagargrowthmedium.
Streptococcussanguinis
Normalfloraofthemouthandthroat.LancefieldgroupH,alphahemolysis.Optochinresistant.Maycause infections inhumans.
Nonmotile.Cocci;chains.Grampositivereaction.Aerobic. Incubationtemperature37°C.Bloodagargrowthmedium.
Streptococcusviridans
Isolatedfromhumansaliva,sputum,and intestine.NoLancefieldgroupshown,alphahemolysis.Forms longchains inbroth
cultures.Nonmotile.Cocci;chains.Grampositivereaction.Aerobic. Incubationtemperature37°C.Bloodagargrowthmedium.
Streptomycesalbus
Isolatedfromstraw.Slightanti-bacterialactivity.Nonmotile.Withcoenocytichyphae.Grampositivereaction.Aerobic. Incubation
temperature37°C.Yeastmaltextractagargrowthmedium.
Streptomycesgriseus
Producesstreptomycin.Nonmotile.Branchingmyceliumandcoenocytichyphae.Grampositivereaction.Aero-
bic. Incubationtemperature25°C.Yeastmaltextractagargrowthmedium.
Vibriofischeri
Found inseawaterandmarineanimals.Youngcultures luminescent.Motile.Rods;curvedandflagellate.Gramnegativereaction.
Aerobic. Incubationtemperature25°C.Photobacteriumagargrowthmedium.
Streptococcuspyogenes
Streptococcusagalactiae
Isolatedfrommilkanduddersofcows.LacefieldgroupB,betahemolysis.Causes infectionofthefemalegenitaltract,maternal
septicemia,neonatal infections.Camptestpositive;resistanttobacitracin.Nonmotile.Cocci;tetradsorchains.Grampositivereac-
tion.Aerobic. Incubationtemperature37°C.Bloodagargrowthmedium.
Streptococcusgallolyticus
Isolatedfromthealimentarytractofcows.LancefieldgroupD,gammahemolysis.Maybepathogenic inhumans.Nonmotile.
Cocci;moderatechains.Grampositivereaction.Aerobic. Incubationtemperature37°C.Bloodagargrowthmedium.
Staphylococcussaprophyticus
Growsondeadtissues.Novobiocinresistant(
S.epidermidis
and
S.aureus
novobiocinsensitive).Coagulasenegative,catalaseposi-
tive.Nonmotile.Cocci;grape-likeclusters.Grampositivereaction.Aerobic. Incubationtemperature37°C.Trypticsoyagargrowth
medium.
Streptomycesviolaceus
Producesavioletcolorthatdiffusesthroughagargrowthmedium.Anti–bacterial,anti–fungal,anti–viralactivities.Nonmotile.
Branchingvegetativemycelium.Grampositivereaction.Aerobic . Incubationtemperature25°C.Sporulationagar.
Key:
Pathogen
n
PlantPathogen
Seethe introductionpageofMoneraforanexplanationofpathogens.
I...,127,128,129,130,131,132,133,134,135,136 138,139,140,141,142,143,144,145,146,147,...1012
Powered by FlippingBook