Ward's Biology 2016-2017 - page 136
Monera
|
LivingMaterials
136
Freeze-DriedCulture
withTransfer
Name
TubeCulture
andGrowthMedia
$11.19
$19.19
EubacteriaCultures(cont.)
Salmonellatyphimurium
,
AmesTestStrain
Amesteststrainfordetectingcarcinogens.Causes infection inhumans.Rods.Motile.Gramnegativereaction.Aerobic. Incubation
temperature37°C.Nutrientagargrowthmedium.
Sarcinaaurantiaca
Producesorangepigments.Nonmotile.Cocci;tetradsandpackets.Grampositivereaction.Aerobic. Incubationtemperature25°C.
Trypticsoyagargrowthmedium.
Serratialiquefaciens
Colorless,mostwidelydispersedoftheSerratiaspecies.Motile.Rods.Gramnegativereaction.Aerobic. Incubationtemperature
30°C.Nutrientagargrowthmedium.
SerratiamarcescensD1
Isolatedfromsoil.Chromogenesis—producesredpigments.Mutatestocolorlessat37°C.Motile.Rods.Gramnegativereaction.
Aerobic. Incubationtemperature25°C.Trypticsoyagargrowthmedium.
Serratiamarcescens933
Colorlessmutant;producesaredpigmentwhengrown inconjunctionwith
S.marcescens
WCF.Motile.Rods.Gramnegativereac-
tion.Aerobic. Incubationtemperature25°C.Nutrientagargrowthmedium.
SerratiamarcescensWCF
Colorlessmutant.Can induceredpigmentformation in
S.marcescens
933.Motile.Rods.Gramnegativereaction.Aerobic. Incuba-
tiontemperature25°C.Nutrientagargrowthmedium.
Shigellaflexneri
Fromsewageandhuman intestine.Causesdysentery.Nonmotile.Rods.Gramnegativereaction.Aerobic. Incubationtemperature
37°C.Nutrientagargrowthmedium.
Shigellasonnei
Isolatedfromhuman intestine.Lactosepositiveandsucrosepositiveafterafewdays’growth.Causesamildformof
dysentery.Nonmotile.Rods.Gramnegativereaction.Facultativelyanaerobic. Incubationtemperature37°C.Nutrientagargrowth
medium.
Sporosarcinaureae
Cocci;cells intetradsorpackets.Motile.Grampositivereaction.Aerobic. Incubationtemperature25°C.Nutrientagargrowth
medium.
Staphylococcusaureus
Causeswound infectionsandtoxicshocksyndrome.Chromogenesis—sometimeshasyellowtoorangepigments.Coagulase
andcatalasepositive.Nonmotile.Cocci;grape-likeclusters.Grampositivereaction.Aerobic. Incubationtemperature37°C.Tryptic
soyagargrowthmedium.
Staphylococcusaureus,
Beta-HemolyticStrain
Betahemolysisonblood.Causeswound infectionsandsepticemia.Coagulaseandcatalasepositive.Nonmotile.Cocci;grape-like
clusters.Grampositivereaction.Aerobic. Incubationtemperature37°C.Trypticsoyagargrowthmedium.
Staphylococcusaureus,
CoagulaseNegative
Relatedto
S.simulans.
Chromogenesis—yelloworgoldenpigmentproduced. Coagulasenegative,catalasepositive.Nonmotile.
Cocci;grape-likeclusters.Grampositivereaction.Aerobic. Incubationtemperature37°C.Trypticsoyagargrowthmedium.
Staphylococcusepidermidis
Normalfloraofhumanskin.Coagulasenegative,catalasepositive.Nonmotile.Cocci;grape-likeclusters.Grampositivereaction.
Aerobic. Incubationtemperature37°C.Trypticsoyagargrowthmedium.
Shigella flexneri
Staphylococcusepidermidis
Bergey’sManualofDeterminativeBacteriology:9thEdition
JohnG.Holt
etal
Areferencetoaid inthe identificationofunknownbacteria, itrepresentsallofthebacteriathathavebeenculturedand
providestaxonomickeystodistinguishbetweenunknowncultures.Softcover,787pages.
$195.00
I...,126,127,128,129,130,131,132,133,134,135 137,138,139,140,141,142,143,144,145,146,...1012
Powered by FlippingBook