Safety Catalogue - page 9
3
Head Protection
Helmets
Description
Colour
Pk
Cat. No.
Vented, without ratchet (EU version)
Safety helmet, vented, without ratchet, EU
Blue
1
Green
1
Black
1
Red
1
White
1
Yellow
1
Vented, without ratchet (UK version)
Safety helmet, vented, without ratchet, UK
Blue
1
Green
1
Black
1
Red
1
White
1
Yellow
1
Description
Colour
Pk
Cat. No.
Unvented, with ratchet (EU version)
Safety helmet, unvented, with ratchet, EU
Blue
1
Green
1
White
1
Yellow
1
Unvented, with ratchet (UK version)
Safety helmet, unvented, with ratchet, UK
Blue
1
Green
1
White
1
Yellow
1
Description
Colour
Pk
Cat. No.
Unvented (EU version)
Safety helmet, unvented, EU
Blue
1
Green
1
Orange
1
Red
1
White
1
Yellow
1
Unvented (UK version)
Safety helmet, unvented, UK
Blue
1
Green
1
Orange
1
Red
1
White
1
Yellow
1
Safety helmets, Concept Roofer
Safety helmets, Eco 1100
Centurion Safety
Centurion Safety
The Concept Roofer is a light, strong and comfortable helmet made from ABS.
Protective helmet with helmet shell made from high density polyethylene (HDPE). For use in
construction and industry.
• With premium ratchet headband ensuring secure fit
• Six point terylene webbing cradle
• Hydro-flock sweatband
• With six point LDPE inner cradle
• Front sweatband made from brushed nylon fabric
• Ear protection, face protection shield and chin strap available as accessories
Size: 520 – 630 mm
Size: 510 – 630 mm
Weight: 340 g
Weight: 375 g
EN 397
EN 397, 440 V, –10 °C, lateral deformation (LD), molten metal (MM)
1,2,3,4,5,6,7,8 10,11,12,13,14,15,16,17,18,19,...526
Powered by FlippingBook