Safety Catalogue - page 8
Head Protection
Helmets
2
Standards
EN 397
Helmets for general use in industry to protect the wearer against falling objects and consequential brain injury and/or skull
fracture.
EN 812
Bump caps protect the wearer against the effects of striking the head/scalp against hard or sharp stationary objects. Not
intended to provide protection against effects of falling, thrown objects or moving suspended loads.
Markings
All safety helmets and bump caps sold in Europe are certified as European Personal Protective Equipment and have the
following symbols as a guarantee of quality:
CE mark
Date of manufacture (day/month/year)
Product mould number
Shift number
Material and properties
Size of helmet
Maintenance
Clean your helmet regularly with soap, water and a soft cloth.
Store out of direct sunlight (between 5 and 25 °C).
Keep your helmet away from chemicals like paint and solvents.
Inspect the quality of your helmet regularly and replace if necessary.
Replace your helmet once every 5 years.
The helmet harness must be replaced every year.
European standards for head protection
Description
Colour
Pk
Cat. No.
Unvented, without ratchet (EU version)
Safety helmet, unvented, without ratchet, EU
Blue
1
Black
1
Red
1
White
1
Yellow
1
Unvented, without ratchet (UK version)
Safety helmet, unvented, without ratchet, UK
Blue
1
Red
1
White
1
Yellow
1
Vision protective helmets
Safety helmets, Concept
Centurion Safety
Centurion Safety
Lightweight, comfortable and modern protective helmet with permanently integrated, hinged
face shield that protects against eye injuries. Helmet shell made from impact resistant ABS
plastic.
Lightweight, comfortable and modern protective helmet made from ABS (acrylonitrile
butadiene styrene) with large forehead area featuring an imprint for the company logo.
• Prescription spectacles can be worn comfortably under the shield
• Six point terylene inner cradle
• Front sweatband made from hydro-flock
• With increased side rigidity (additional lateral deformity test)
• Easily replaceable, transparent protective visor made from polycarbonate, weight: 30 g
• With standard slip headband or twist ratchet
• With extended nape area
• Six point terylene inner cradle
• Highly absorbent hydro-flock sweatband
• With increased side rigidity (additional lateral deformity test)
• Ventilation through wide ventilation slots
• Without ratchet, with full peak
Size: 51 – 63 cm, 52 – 63 cm with ratchet
Size: 510 – 630 mm
Weight: 495 g including inner cradle
Weight: 325 g
Helmet: EN 397; Integrated visor: EN 166
EN 397
1,2,3,4,5,6,7 9,10,11,12,13,14,15,16,17,18,...526
Powered by FlippingBook