Safety Catalogue - page 41
35
Hearing Protection
Earplugs
Earplugs
Description
Colour
Size
Pk
Cat. No.
Soft earplugs, without cord, packed in pillow
packs
Yellow
U*
200 Pair
Soft earplugs, without cord, 200 pairs per box Yellow
U*
200 Pair
Soft earplugs, with cord, 200 pairs per box
Yellow
U*
200 Pair
Pillow case
Disposable earplugs, E-A-R™ Classic™ Soft
3M
Foam, without cord
Description
Colour
Size
Pk
Cat. No.
Earplugs, without cord, pillow pack
Yellow
S
250 Pair
Earplugs, with blue polymer cord
Yellow
U*
200 Pair
Earplugs, without cord, small bag of 5 pairs
Yellow
U*
250 Pair
Earplugs, without cord, pillow pack
Yellow
U*
250 Pair
One-Touch™ dispenser for disposable E-A-R™ earplugs
Disposable earplugs, E-A-R™ Classic™
These earplugs feature a unique PU foam combination which warms to the body temperature
and becomes softer, thus allowing the earplug to mould to the individuals ear canal shape.
The earplugs therefore provide even more comfort to the end user than the other Classic™
earplugs.
3M
Foam, with or without cord
• Provide an efficient seal due to the moulding of the earplug to the ear canal
• Textured surface provides greater friction which improves grip and helps maintain an
effective seal
• Earplug is resistant to moisture, making it ideal for hot and humid working environments
The 3M™ One-Touch™ dispenser system is an innovative solution that can be used across a range of 3M™ E-A-R™ earplugs. Easy to use
with a simple twist motion, the innovative 'no waste funnel' ensures plugs drop directly into the palm, dispensing disposable, uncorded
earplugs every time.
These roll-down earplugs are made from a soft, energy absorbing polymer foam which
provides excellent hearing protection and all-day comfort. Ideal for use where noise is a
problem either in work or leisure activities. Also available in a small version for users with
narrow ear canals, and in a corded version.
SNR: 36 dB
• Versatile design includes tabletop or wall-mount options for convenient dispensing
• Unobstructed access from front or either side permits one handed operation by a clockwise or anticlockwise motion
• Firm material with slow recovery time helps to achieve good fit
• Sweat and humidity resistant which improves comfort
• Very economical as the earplugs can be cleaned up to three times
EN 352-2
Delivery information:
The dispenser is supplied without earplugs. Please order the refill bottles separately.
*One size fits all
SNR: 28 dB
EN 352-2
Packaging:
Either single pairs in pillow packs, 5 pairs in small bags or 500 pairs in dispenser
box.
*One size fits all
1...,31,32,33,34,35,36,37,38,39,40 42,43,44,45,46,47,48,49,50,51,...526
Powered by FlippingBook