Safety Catalogue - page 360
Fall Protection
Harness
354
EN 364
Test methods for personal equipment against falls from a height
EN 365
General requirements for instructions for use and for marking
EN 341
Descender devices
EN 353-1
Part 1: Guided type fall arresters including a rigid anchor line
EN 353-2
Part 2: Guided type fall arresters including a flexible anchor line
EN 354
Lanyards
EN 355
Energy absorbers
EN 358
Belts for work positioning and restraint and work positioning lanyards
EN 360
Retractable type fall arresters
EN 361
Full body harnesses
EN 362
Connectors
EN 363
Fall arrest systems
EN 795
Anchor devices - requirements and testing
EN 813
Sit harnesses
Fall protection norms: Personal protective equipment against falls from a height
Type
Size
Pk
Cat. No.
2-point
S/M
1
2-point
L/XL
1
Accessories
Description
Pk
Cat. No.
Holder for mobile phones/radios, with snap-on connector, black
1
Large tool bag with hammer loops, 190×290 mm
1
Safety harness, Revolution Comfort R2
Miller by Honeywell
Two-point safety harness for use with fall protection equipment and a suitable anchorage point. The webbing is made from DuraFlex®, a highly
elastic stretch fabric. Thanks to its unique technology and versatility, the harness is ideally suited for scaffolding, steel and mast construction
applications, as well as for general use and maintenance.
• Safety loop at the back (D-ring) and the front (webbing loops)
• PivotLink connection at waist height for increased mobility and comfort thanks to optimum ergonomics when bending and stretching
• Single-handed adjustment thanks to cam buckles and harness clips; automatic quick-lock mechanisms
• ErgoArmor™ padded back shield for added comfort
• Integrated accessory system: Pockets for tools and accessories can easily be fastened to or removed from the modular attachment system
EN 361
1...,350,351,352,353,354,355,356,357,358,359 361,362,363,364,365,366,367,368,369,370,...526
Powered by FlippingBook