Safety Catalogue - page 290
Workwear
High Visiblility
284
EN 471 (2-2)
EN 61482-1-2 (class 1), EN ISO 11612 (A1 A2 B1 C1), EN 1149-5,
EN 13034 type PB (6)
Colour
Clothing size
Pk
Cat. No.
Hi-Vis Yellow
U*
1
Hi-Vis Orange
U*
1
Colour
Clothing size
Pk
Cat. No.
Blue/Hi-Vis yellow
XS
1
Blue/Hi-Vis yellow
S
1
Blue/Hi-Vis yellow
M
1
Blue/Hi-Vis yellow
L
1
Blue/Hi-Vis yellow
XL
1
Blue/Hi-Vis yellow
XXL
1
Blue/Hi-Vis yellow
3XL
1
High visibility road waistcoat, Essential
Flame-retardant jacket, Flamestat
Kwintet
Kwintet
Waistcoat with two reflective strips, made from 100% polyester.
This jacket is made from 54% modacrylic, 44% cotton and 2% anti-static fibres and is resistant
to dirt, oil and water. The jacket also offers basic protection against heat, open flames and
arcing faults, and in addition is certified for work in areas where there is the risk of electrostatic
discharge. Basic protection against chemicals is also provided.
• With rip-tape tab
• Packed in a practical pouch
• Concealed zip and buttons up to the top of the collar
• Adjustable bottom edge and cuffs
• Breast pockets with flap and concealed press stud, one with tool loop
• Two concealed breast pockets with zip, one with opening and loop for earphones
• Concealed button and loop for work knife
• Two front pockets, inner pocket with button
Weight: 110 g/m²
Weight: 320 g/m²
*One size fits all
1...,280,281,282,283,284,285,286,287,288,289 291,292,293,294,295,296,297,298,299,300,...526
Powered by FlippingBook