Safety Catalogue - page 25
19
Eye & Face Protection
Spectacles & Goggles
Type
Lenses
Colour
Pk
Cat. No.
Anti-UV (2C-1.2), anti-scratch, anti-fog, anti-static
Cear PC
Orange/Blue
1
Anti-UV (2C-1.2), anti-scratch, anti-fog
Clear PC
Blue/Grey
1
Anti-UV (2C-1.2), anti-fog (high performance)
Clear PC
Green/White
1
Type
Lenses
Colour
Pk
Cat. No.
Pheos
Anti-UV, anti-scratch, anti-fog
Clear PC
Light Grey/Grey
1
Anti-UV, anti-scratch, anti-fog
Clear PC
Black/Green
1
Anti-UV, anti-scratch, anti-fog
Amber PC
Black/Yellow
1
Anti-UV, anti-scratch, anti-fog
Clear PC, with sunglare filter
Light Grey/Grey
1
Type
Lenses
Colour
Pk
Cat. No.
Anti-UV (2-1.2), anti-scratch
Clear PC
Orange/Black
1
Anti-UV (2-1.2), anti-scratch, anti-fog
Clear PC
Orange/Black
1
Anti-UV (5-3.1), anti-scratch, anti-fog
Bronze PC
Orange/Black
1
Anti-UV (5-1.7), anti-scratch, anti-fog
I/O mirrored PC
Orange/Black
1
Anti-UV (5-3.1), anti-scratch, anti-fog
Red mirrored PC
Orange/Black
1
Anti-UV (5-3.1), anti-scratch, anti-fog
Mirrored PC
Blue/Grey
1
Safety spectacles, I-3
Description
Type
Lenses
Colour
Pk
Cat. No.
Pheos, for narrow heads
Small version for narrow heads
Anti-UV, anti-scratch, anti-fog
Clear PC
White-Green
1
Small version for narrow heads
Anti-UV, anti-scratch, anti-fog
Clear PC
Anthracite
1
Pheos Blue, eco-friendly
Eco-friendly, based on natural raw materials (organic)
Anti-UV, anti-scratch, anti-fog
Clear PC
Beige/Blue
1
Safety spectacles, Solus™
Uvex
Safety spectacles, Pheos
3M
Single lens
Uvex
Dual lens
These innovative safety spectacles are designed for a perfect fit.
Single lens
Extremely stylish and lightweight eyewear available with blue lens or with a light flash mirror
coating. It removes eye stress and reduces eye fatigue often occurring in monochromatic
(single colour) yellow lit environments. Perfect in work environments where there is strong
glare from bright lighting (outdoor, semiconductor fabrication and similar industries where
there is medium to high levels of yellow incandescent/fluorescent lamps).
• Flexible side arms with variable inclination
• Integrated Softlex zone ensures comfort for all head sizes
• Soft nosepiece, earpieces and Softflex zone at temples ensure a comfortable and secure fit
The supravision HC-AF coating technology makes the uvex Pheos lens highly scratch
resistant on the outside and permanently anti-fog on the inside.
• Lens curve optimises the fitting, field of vision and coverage of the eyes
• Several lens options are available
EN 166, EN 170
• Provides 100% UV protection
• Integral brow guard for additional coverage
• Enhanced ventilation
• High performance hinges
• PPE compatibility (earmuffs, safety helmets and bump caps)
EN 166, EN 170
EN 166, EN 170
Delivery information:
Supplied with pouch.
1...,15,16,17,18,19,20,21,22,23,24 26,27,28,29,30,31,32,33,34,35,...526
Powered by FlippingBook