Safety Catalogue - page 22
Eye & Face Protection
Spectacles & Goggles
16
Type
Lenses
Colour
Pk
Cat. No.
Anti-UV, anti-fog
Clear PC
Red/Silver-Grey
1
Anti-UV, anti-scratch
Clear PC
Black
1
Anti-UV, anti-scratch
TSR smoked PC
Red/Silver-Grey
1
Type
Lenses
Colour
Pk
Cat. No.
Anti-UV, anti-scratch
Clear PC
Black
1
Anti-UV, anti-scratch
HDL-Yellow PC
Black
1
Anti-UV, anti-scratch
I/O mirrored PC
Black
1
Anti-UV, anti-scratch
Mirrored PC
Black
1
Anti-UV, anti-scratch
TSR-grey PC
Black
1
Anti-UV, anti-scratch, anti-fog
TSR-grey PC
Black
1
Insert with +1.5 dpt.
1
Insert with +2.5 dpt
1
Type
Lenses
Colour
Pk
Cat. No.
Anti-fog, anti-scratch
TSR-grey PC
Black
1
Anti-scratch
TSR-grey PC
Black
1
Anti-fog, anti-scratch
Clear PC
Black
1
Anti-scratch
Clear PC
Black
1
Type
Lenses
Colour
Pk
Cat. No.
Anti-UV (5-3.1), anti-scratch, anti-fog TPR smoke PC
Black
1
Anti-UV (2-1.2), anti-scratch, anti-fog
TPR clear PC
Black
1
Safety spectacles, Universal
Safety spectacles, Ignite
Safety spectacles, Protégé™
Safety spectacles, FitLogic®
Bollé Safety
Honeywell
Honeywell
Honeywell
Single lens
Single lens
Single lens
Single lens
Extremely comfortable fit thanks to the flexible, lightweight bridge and the pivoting temples.
These spectacles allow for general use and protection against impact. Suitable for applications
in mechanical engineering.
These spectacles are suitable for indoor use against most commonly encountered hazards.
These spectacles are suitable for applications such as utilities, light and heavy manufacturing,
biotech, ship building, construction and automotive manufacturing.
• Anti-scratch, anti-fog coatings
• Upper and lower protection
• Non slip B-Flex nose bridge
• Pivoting, co-injected straight temples
• Protection against impact from −5 to +55 °C
• Flexible, expandable and durable
• Lightweight and comfortable
• Superior coverage: Close-to-the-face protection
• Lightweight frameless design: Comfortable fit
• Adjustable nosepiece reduces slippage
• Lightweight design and stylish sunglass look
• Flexible temples: Improved comfort
• Soft nose bridge
Weight: 26 g
Weight: 26 g
EN 166, EN 170
Weight: 27,5 g
EN 160, EN 170
EN 166, EN 170
1...,12,13,14,15,16,17,18,19,20,21 23,24,25,26,27,28,29,30,31,32,...526
Powered by FlippingBook