Safety Catalogue - page 21
15
Eye & Face Protection
Spectacles & Goggles
Type
Lenses
Colour
Pk
Cat. No.
Anti-UV (5-3.1), anti-scratch, anti-fog
Blue flash
polarised PC
Black
1
Smoke PC
Black
1
Type
Lenses
Colour
Pk
Cat. No.
Anti-UV (5-3.1), anti-scratch, anti-fog
Smoke PC
Brown
1
Smoke PC
Black
1
Amber PC
Brown
1
Type
Lenses
Colour
Pk
Cat. No.
Anti-UV, anti-scratch, anti-fog
Smoke PC
Black
1
Anti-scratch, anti-fog
Clear PC
Transparent
1
Type
Lenses
Colour
Pk
Cat. No.
Anti-scratch, anti-fog
Clear PLA and biodegradable plastic
Black
1
Safety spectacles, Solis B-Green
Safety spectacles, Tracker
Safety spectacles, Voodoo
Safety spectacles, Hustler
Bollé Safety
Bollé Safety
Bollé Safety
Bollé Safety
Dual lens, environmentally friendly
Dual lens
Dual lens
Dual lens
Solis B-green is made from PLA and renewable materials. The frame contains 40% biocarbon
and the manufacturing and the recycling of the materials both produce less greenhouse gas.
These sporty design safety spectacles feature a durable PC frame, a smoke PC dual lens,
straight temples with upper and lower protection and an adjustable headband.
Ultra-wrap and sporty, this model perfectly combines comfort, protection and design. Available
in smoke, brown and polarised brown.
Ultra-wrap and sporty, these safety spectacles perfectly combine comfort, protection and
design. Available in smoke and blue flash polarised version.
• OK-Biobased certified by the Belgian laboratory Vincotte
• Screwless and lightweight frame
• Anti-scratch and anti-fog coating for improved safety
• Side protection
• Non slip bridge
• Anti-scratch, anti-fog coatings
• Non slip temples
• Interchangeable foam reinforcement with indirect ventilation
• Adjustable head strap and rubber arm tips provide a secure fit and greater protection
• ALS anti-fog/anti-scratch coating on both sides of clear and smoke lenses
• Rubberised eye seal provides exceptional protection against dust and other particles
• Side protection
• Non slip bridge
• Anti scratch, anti fog coatings
• Non slip temples
Weight: 18 g
Weight: 29 g
Weight: 52 g
Weight: 25 g
EN 166, EN 170
EN 166
EN 166, EN 172
EN 160, EN 170, EN 172
Delivery information:
Supplied in a box made from recycled cardboard.
Ordering information:
Supplied with microfibre pouch.
1...,11,12,13,14,15,16,17,18,19,20 22,23,24,25,26,27,28,29,30,31,...526
Powered by FlippingBook