Safety Catalogue - page 19
13
Eye & Face Protection
Spectacles & Goggles
Type
Lenses
Colour
Pk
Cat. No.
With headband: Anti-UV (2-1.2), anti-
scratch
Clear PC
Transparent
1
Type
Lenses
Colour
Pk
Cat. No.
Anti-UV (5-3.1/ 100% UV), anti-scratch,
anti-fog, anti-static
Smoke PC
Black
1
Anti-UV, anti-scratch, anti-fog, anti-
static
Clear PC
Black
1
Type
Lenses
Colour
Pk
Cat. No.
Anti-UV 2-1.2, anti-scratch, anti-fog
Clear PC
Grey
1
Anti-UV 5-1.4, anti-scratch
Orange PC
Grey
1
Anti-UV 5-1.4, anti-scratch, anti-fog
Smoke PC
Grey
1
Safety spectacles, Slam
Safety spectacles, B-Line
Safety spectacles, Contour
Bollé Safety
Bollé Safety
Bollé Safety
Dual lens
Single lens
Dual lens
Modern design spectaclesl with extra-easy fitting and comfort.
Solidity, protection, design, comfort and lightness make the Contour spectacles a unique
model.
Safety spectacles, Contour Metal
Spectacles offering a panoramic field of vision. The frame is manufactured from nylon.
• Panoramic visual field
• Anti-scratch, anti-fog, anti-static coatings
• Ultra enveloping
• Comfort temples
• Integrated side shields
• With headband
• Non slip TIPGRIP temples
• Non slip bridge
Bollé Safety
Weight: 23 g
Weight: 21 g
Weight: 30 g
Dual lens
EN 166, EN 170 (clear lens), EN 172 (smoke lens)
EN 166, EN 170
EN 166, (EN 170, EN 172)
Contour spectacles feature great innovative design.
Delivery information:
Supplied with adjustable cord.
Delivery information:
Supplied with microfibre pouch.
• 160% flex temples
• Non slip bridge
• Increased upper protection
Weight: 38 g
EN 166, EN 170 (clear version), EN 172 (orange version)
Delivery information:
Supplied with microfibre pouch.
1...,9,10,11,12,13,14,15,16,17,18 20,21,22,23,24,25,26,27,28,29,...526
Powered by FlippingBook