Safety Catalogue - page 17
11
Eye & Face Protection
Spectacles & Goggles
Type
Lenses
Colour
Pk
Cat. No.
Anti-UV (2-1.2), anti-scratch, anti-fog
Clear PC
Transparent
1
Type
Lenses
Colour
Pk
Cat. No.
Anti-UV, anti-scratch, anti-fog
Clear PC
Red
1
Anti-UV, anti-scratch
Indoor/outdoor PC
Amber
1
Type
Lenses
Colour
Pk
Cat. No.
Anti-UV (2-1.2), anti-scratch, anti-fog
Clear PC
Transparent
1
Safety spectacles, Dublin
Safety spectacles, Milano
Safety spectacles, Vienna
Single lens
Single lens
Single lens
Spectacles in a fashionable design, ideal for women due to low cut profile. Provide
excellent field of vision. Ideal for working with laboratory UV lamps.
Lightweight spectacles with clear polycarbonate lens.
Type
Lenses
Colour
Pk
Cat. No.
Anti-UV, anti-scratch, anti-fog
Clear PC
Transparent
1
Anti-UV, anti-scratch, anti-fog
Smoke PC
Black
1
Lightweight safety spectacles with clear polycarbonate lens.
Safety spectacles, Berlin
• High UV protection (UV 254 tested)
• Slim temples, fashionable design
• Anti-scratch/anti-fog coating
• Moulded brow guard
• Soft nose pad
• Soft rubber temple pads
• Anti-scratch/anti-fog coating
• With pantoscopic adjustment for angle modification
• Moulded brow guard
• Adjustable temple length
Weight: 25.5 g
Weight: 27 g
Single lens
Weight: 26 g
The innovative design of these spectacles provides an excellent field of vision. Ideal
for working with laboratory UV lamps.
EN 166, EN 170
EN 166, EN 170; 1 FT
EN 166, EN 170; 1 FT
• High UV protection (UV 254 tested)
• Moulded brow guard
• Soft temple tips, made from two materials
• Adjustable temples for a perfect fit
Weight: 29 g
EN 166, EN 170 (EN 172 -
1...,7,8,9,10,11,12,13,14,15,16 18,19,20,21,22,23,24,25,26,27,...526
Powered by FlippingBook