Safety Catalogue - page 13
7
Head Protection
Accessories
Type
L×W (mm)
Colour
Pk
Cat. No.
With pictogram "Face shield"
216×196
Blue
1
With pictogram "Eye protection"
216×196
Blue
1
With pictogram "Ear protection"
216×196
Blue
1
With pictogram "Respirator"
216×196
Blue
1
With pictogram "Safety helmet'
216×196
Blue
1
With pictogram "Protection frocks"
216×196
Blue
1
Wall Hooks, SecuPoint
Description
Colour
W×D×H (mm)
Pk
Cat. No.
Mini
Eye protection
Blue
236×120×120
1
Eye protection
Transparent
236×120×120
1
Hair nets
Blue
236×120×120
1
Hair nets
Transparent
236×120×120
1
Hand protection
Blue
236×120×120
1
Hand protection
Transparent
236×120×120
1
Midi
Ear protection
Blue
236×125×225
1
Ear protection
Transparent
236×125×225
1
Respirator
Blue
236×125×225
1
Respirator
Transparent
236×125×225
1
Hair nets
Blue
236×125×225
1
Hair nets
Transparent
236×125×225
1
Maxi
Helmet
Blue
236×200×315
1
Helmet and eye protection
Blue
236×200×315
1
Strong ABS
Dispensing boxes
• Easy installation by screw attachment
ABS
These containers are designed for storing personal safety apparatus in the workplace.
They can be mounted using adhesive fittings or can be screwed to the wall.
• Protection against dirt, dust and dampness
• Patented opening allows for quick removal of the PPE in case of emergency
• Available in three sizes
1...,3,4,5,6,7,8,9,10,11,12 14,15,16,17,18,19,20,21,22,23,...526
Powered by FlippingBook