Safety Catalogue - page 11
5
Head Protection
Bump Caps
Colour
Pk
Cat. No.
Hi-Vis Orange
1
Hi-Vis Yellow
1
Colour
Pk
Cat. No.
Blue
1
Green
1
Orange
1
Red
1
White
1
Yellow
1
High visibility bump caps, CoolCap
Bump caps, Bump Cap™ S19
Centurion Safety
Centurion Safety
This polyester bump cap is fitted over an ABS liner to provide lightweight head protection
where high impact resistance is not essential, but where there is a risk of minor bumps or
scrapes to the head. Suitable applications include automotive, airlines, transportation,
shipping, railways and engineering.
This polyethylene cap is ideal for use in the food industry and in other areas where textile
clothing materials are not allowed for reasons of hygiene and where sterility is required.
• Shell made of lightweight ABS
• Breathable thanks to sleek three panel design with improved side ventilation
• Removable foam side inserts provide 45% more shock absorbency than standard ABS liner
• Fluorescent colours for improved visibility
• Sweatband made from PVC, LDPE headband and interiors are washable and replaceable
• Lightweight and durable
• Outer shell is easily washable
For head size 54 – 59 cm
Size: 54 – 59 cm
Weight: 155 g
Weight: 195 g
EN 812
BS EN 812: 1997; amendment no. 1 (2001) with CE marking
1...,2,3,4,5,6,7,8,9,10 12,13,14,15,16,17,18,19,20,21,...526
Powered by FlippingBook