Safety Catalogue - page 100
Gloves
Disposable Gloves - Nitrile
94
Type
Size
Pk
Cat. No.
Powdered, textured fingertips, Length: 240 mm
92-500
6,5 - 7
100
92-500
7,5 - 8
100
92-500
8,5 - 9
100
92-500
9,5 - 10
100
Powder-free, smooth finish, Length: 240 mm
92-600
6,5 - 7
100
Gloves, Touch N Tuff® 92-500, 92-600 and 92-605
Size
Pk
Cat. No.
S
100
M
100
L
100
XL
90
Length (mm)
Size
Pk
Cat. No.
240
XS
100
240
S
100
240
M
100
240
L
100
240
XL
90
300
XS
100
300
S
100
300
M
100
300
L
100
300
XL
90
Ansell
Gloves, Nitrile
Gloves, Nitrile Light
Nitrile, non sterile, powdered or powder-free, smooth finish or textured fingertips
Nitrile, non sterile, powder-free, textured finish
Nitrile, non sterile, powder-free, textured fingertips
Gloves feature rolled cuffs for wrist protection and to keep the glove in place. The glove is
made of puncture resistant nitrile which offers valuable protection against accidental splashes.
For laboratory use in pharmaceutical, industrial and medical applications, and in the
food industry. These ambidextrous gloves provide extra protection, particularly for
sensitive skin.
• 100% nitrile; no waxes, silicone or plasticisers; ideal for sensitive skin
• Three times the puncture resistance of latex or PVC
• Light, cool and durable
• Designed for maximum freedom of movement and minimum hand fatigue
Light, ambidextrous and durable gloves that are ideal for food handling, catering and
kitchen work, medical applications and other complex risk tasks. The thin nitrile gloves
are free from phthalates, softeners and allergenic latex proteins.
• Highly resistant to chemical solvents and fats
• Excellent grip, especially when holding wet objects
• Designed especially for latex allergy sufferers
• Low thickness and textured fingertips for excellent grip
• Particularly skin friendly for users with latex allergy
• Rolled cuffs minimise the risk of tearing
• Suitable for food contact
Thickness: 0,12 mm
Length: 240 mm
Length: 240 or 300 mm
Colour: Green
Thickness: 0,25 mm (double wall)
Thickness: 0,18 mm (double wall)
EN 374, Cat. III; AQL 1.5
Colour: Blue
Colour: Blue
Tested according EN 1149-1 and 1149-3 (anti-static properties)
EN 374 (Cat. III), EN 455, AQL 1,5
EN 374 (Cat. III), EN 455, AQL 1,5
1...,90,91,92,93,94,95,96,97,98,99 101,102,103,104,105,106,107,108,109,110,...526
Powered by FlippingBook