Safety Catalogue - page 10
Head Protection
Helmets
4
Description
Colour
Pk
Cat. No.
Unvented (EU version)
Safety helmet, unvented, EU
Blue
1
Green
1
Black
1
Orange
1
Red
1
White
1
Yellow
1
Hi-Vis Orange
1
Hi-Vis Yellow
1
Unvented (UK version)
Safety helmet, unvented, UK
Blue
1
Green
1
Black
1
Orange
1
Red
1
White
1
Yellow
1
Vented (EU version)
Safety helmet, vented, EU
Blue
1
Green
1
Orange
1
Red
1
White
1
Yellow
1
Description
Colour
Pk
Cat. No.
Safety helmet, heat resistant
White
1
Safety helmet, heat resistant
Yellow
1
Eco 1125 protective helmets
Safety helmets, Vulcan
Centurion Safety
Centurion Safety
Impact resistant protective helmet with helmet shell made from high density polyethylene
(HDPE). Suitable for the construction industry.
Heat resistant thermoset helmet made from glass fibre reinforced polyester resin. Ideal for use
in industries that generate heat.
• With six point terylene inner cradle
• Front sweatband made from brushed nylon fabric
• Ear protection, face protection shield and chin strap available as accessories
• With six point terylene inner cradle
• Front sweatband made from brushed nylon fabric
• Temperature resistant up to 200 °C
• Ear protection, face protection shield and chin strap available as accessories
Size: 510 – 630 mm
Size: 510 – 630 mm
Weight: 365 g
Weight: 375 g
EN 397 (unvented version); Electrical Resistance and Molten Metal applicable to unvented
shells only.
EN 397, 440 V, –30 °C, lateral deformation (LD), molten metal (MM)
1,2,3,4,5,6,7,8,9 11,12,13,14,15,16,17,18,19,20,...526
Powered by FlippingBook