VWR Healthcare Clinical Catalog 2012-2014 - page 346
PerfeCTa
®
qPCR SuperMixes, Quanta Biosciences
Unique buffer and stabilizers have been specifically optimized to deliver maximum PCR
efficiency, sensitivity, and a robust fluorescent signal with TaqMan
®
or TaqMan
®
MGB probe
chemistry. The enhanced specificity of this supermix suppresses cross-reactivity between
homologous sequences, improving detection and discrimination in SNP applications. A key
component of this supermix is AccuStart
Taq DNA polymerase with monoclonal antibodies
that bind to the polymerase and keep it inactive prior to the initial PCR denaturation step.
Upon heat activation (two minutes at 95°C), the antibodies denature irreversibly, releasing
fully active, unmodified Taq DNA polymerase. This enables specific and efficient primer
extension with the convenience of room temperature reaction assembly.
Supermixes are available and optimized for all real-time PCR instrument platforms including
those requiring normalization with ROX
dyes. Supermixes are also available with uracil-
N-glycosylase (UNG) to prevent amplification of carry-over contamination from previous
dU-containing PCRs.
Reactions/Kit
Supplier No.
Cat. No.
Price
SuperMix
100
95050-100
Ea./
177.00
500
95050-500
Ea./
869.00
2000
95050-02K
Ea./
3274.00
SuperMix with Low ROX
100
95052-100
Ea./
177.00
500
95052-500
Ea./
869.00
2000
95052-02K
Ea./
3274.00
SuperMix with ROX
100
95051-100
Ea./
177.00
500
95051-500
Ea./
869.00
2000
95051-02K
Ea./
3274.00
SuperMix with UNG
100
95064-100
Ea./
186.00
500
95064-500
Ea./
913.00
2000
95064-02K
Ea./
3438.00
SuperMix with UNG and Low ROX
100
95066-100
Ea./
186.00
500
95066-500
Ea./
913.00
2000
95066-02K
Ea./
3438.00
SuperMix with UNG and ROX
100
95065-100
Ea./
186.00
500
95065-500
Ea./
913.00
2000
95065-02K
Ea./
3438.00
1.800.932.5000
|
vwr.com
|
VWR International
|
333
Molecular Diagnostics
PCR Kits
1...,336,337,338,339,340,341,342,343,344,345 347,348,349,350,351,352,353,354,355,356,...955
Powered by FlippingBook