VWR Healthcare Clinical Catalog 2012-2014 - page 343
4-CORE
®
Buffer Pack, Promega
®
Buffer pack contains convenient aliquots of Promega
®
restriction enzyme 10X buffers A, B, C, and D. The majority of Promega
®
restriction enzymes have optimal activ-
ity in one of these four 10X reaction buffers.
Note:
For laboratory use. Outside of the United States, this product is intended for research use only, unless otherwise stated.
Ordering Information:
Pack contains 1mL each of buffers A, B, C, and D.
Description
Size
Supplier No.
Cat. No.
Price
Buffer Pack
4 x 1 mL
R9921
Ea./
10.00
MULTI-CORE
Buffer Pack, Promega
®
Buffer pack contains convenient aliquots of the Promega
®
universal restriction enzyme 10X buffer. Formulated to provide simple buffering conditions for performing
multiple digestions. Many Promega
®
restriction enzymes have between 50% and 100% activity in reactions using this buffer.
Note:
For laboratory use. Outside of the United States, this product is intended for research use only, unless otherwise stated.
Description
Size
Supplier No.
Cat. No.
Price
Buffer Pack
3 x 1 mL
R9991
Ea./
10.00
qScript
cDNA SuperMix, Quanta Biosciences
SuperMix provides a sensitive and easy-to-use solution for two-step qRT-PCR. This 5x concentrated master mix provides all necessary
components (except RNA template) for first-strand synthesis including: buffer, dNTPs, MgCl2, primers, RNase inhibitor protein, qScript
reverse transcriptase, and stabilizers. qScript is a RNase H(+) derivative of MMLV-RT, optimized for reliable cDNA synthesis over a
wide dynamic range of input RNA. The unique blend of oligo (dT) and random primers in the qScript cDNA SuperMix works exception-
ally well with a wide variety of targets. This blend is optimized for the production of targets <1kb in length. qScript cDNA SuperMix
produces excellent results in both real-time and conventional RT-PCR.
Description
Supplier No.
Cat. No.
Price
5 x 96-Well Plates
95048-096
Ea./
2753.00
25 Reactions
95048-025
Ea./
129.00
100 Reactions
95048-100
Ea./
512.00
500 Reactions
95048-500
Ea./
2506.00
qScript
cDNA Synthesis Kits, Quanta Biosciences
Kits are a sensitive and easy-to-use solution for RNA quantification using two step RT-PCR. The qScript
reaction mix provides all
the necessary components for cDNA synthesis except the enzyme and RNA template. The optimized blend of random and oligo(dT)
primers provides robust, consistent, and unbiased first-strand synthesis over a broad range of RNA template concentrations. qScript
reverse transcriptase is a mixture of an engineered MMLV-RT and a ribonuclease inhibitor protein. The simplified reaction procedure is ideally suited for high through-
put expression studies using RT-PCR. The resulting cDNA product is directly compatible with current real-time PCR methods or conventional end-point RT-PCR where
the targets are ≤1kb in length.
Reactions/Kit
Supplier No.
Cat. No.
Price
25
95047-025
Ea./
110.00
100
95047-100
Ea./
366.00
500
95047-500
Ea./
1737.00
Antigen Unmasker, Diagnostic Biosystems
This antigen unmasker is in tablet form and is free from heavy metals. Fixative used on tissue sections to conserve the tissue from autolysis, preserve
tissue structures, and immobilize antigens alters the tissue. Antigens undergo chemical alteration of their primary, secondary and tertiary structures.
Because of changes in the protein-containing epitopes or in neighboring proteins, antigenic sites may be masked. Use of a heavy metal solution in
a microwave can regain the masked antigens significantly. However, heavy metals increase the risk of exposure of lab personnel to lead. Use of this
antigen unmasker can eliminate the risk of heavy metal exposure to lab personnel and also resolve heavy metals handling and disposal problems.
Description
Supplier No.
Cat. No.
Price
Antigen Unmasker,Tablets
DSI 5030
Ea./
85.00
330
|
VWR International
|
vwr.com
|
1.800.932.5000
Molecular Diagnostics
Media and Reagents
1...,333,334,335,336,337,338,339,340,341,342 344,345,346,347,348,349,350,351,352,353,...955
Powered by FlippingBook