VWR Healthcare Clinical Catalog 2012-2014 - page 338
Size
Enzyme
Recognition/Cut Site
Supplier No. Cat. No.
Price
15,000 Units, High Concentration, 40–80 Units/μL Pst I (Blue/White Cloning Qualified)
CTGCAG
GACGTC
R4114
Ea./
131.00
50,000 Units, High Concentration, 40–80 Units/μL Pst I (Blue/White Cloning Qualified)
CTGCAG
GACGTC
R4117
Ea./
267.00
3000 Units
Pst I (Blue/White Cloning Qualified)
CTGCAG
GACGTC
R6111
Ea./
36.00
15,000 Units
Pst I (Blue/White Cloning Qualified)
CTGCAG
GACGTC
R6115
Ea./
131.00
100 Units
Pvu I
CGATCG
GCTAGC
R6321
Ea./
65.00
500 Units
Pvu I
CGATCG
GCTAGC
R6325
Ea./
263.00
1000 Units
Pvu II
CAGCTG
GTCGAC
R6331
Ea./
45.00
5000 Units
Pvu II
CAGCTG
GTCGAC
R6335
Ea./
169.00
5000 Units, High Concentration, 40–80 Units/μL Rsa I
GTAC
CATG
R4374
Ea./
212.00
1000 Units
Rsa I
GTAC
CATG
R6371
Ea./
57.00
5000 Units, High Concentration, 40–80 Units/μL Sac I (EcoICR I) (Blue/White Cloning Qualified)
GAGCTC
CTCGAG
R4064
Ea./
210.00
1000 Units
Sac I (EcoICR I) (Blue/White Cloning Qualified)
GAGCTC
CTCGAG
R6061
Ea./
60.00
5000 Units
Sac I (EcoICR I) (Blue/White Cloning Qualified)
GAGCTC
CTCGAG
R6065
Ea./
209.00
500 Units
Sac II (Blue/White Cloning Qualified)
CCGCGG
GGCGCC
R6221
Ea./
68.00
10,000 Units, High Concentration, 40–80 Units/μL Sal I (Blue/White Cloning and Genome Qualified)
GTCGAC
CAGCTG
R4054
Ea./
255.00
2000 Units
Sal I (Blue/White Cloning and Genome Qualified)
GTCGAC
CAGCTG
R6051
Ea./
71.00
10,000 Units
Sal I (Blue/White Cloning and Genome Qualified)
GTCGAC
CAGCTG
R6055
Ea./
256.00
100 Units
Sau3A I (Dpn I, Mbo I)
GATC
CTAG
R6191
Ea./
46.00
500 Units
Sau3A I (Dpn I, Mbo I)
GATC
CTAG
R6195
Ea./
178.00
5000 Units, High Concentration, 40–80 Units/μL Sca I
AGTACT
TCATGA
R4214
Ea./
251.00
1000 Units
Sca I
AGTACT
TCATGA
R6211
Ea./
70.00
1250 Units, High Concentration, 40–80 Units/μL Sfi I (Blue/White Cloning and Genome Qualified)
GGCCNNNNNGGCC
CCGGNNNNNCCGG
R4394
Ea./
234.00
250 Units
Sfi I (Blue/White Cloning and Genome Qualified)
GGCCNNNNNGGCC
CCGGNNNNNCCGG
R6391
Ea./
59.00
1250 Units
Sgf I
GCGATCGC
CGCTAGCG
R5104
Ea./
296.00
250 Units
Sgf I
GCGATCGC
CGCTAGCG
R7103
Ea./
78.00
1000 Units, High Concentration, 40–80 Units/μL Sin I (Ava II)
GG(A/T)CC
CC(T/A)GG
R4144
Ea./
183.00
200 Units
Sin I (Ava II)
GG(A/T)CC
CC(T/A)GG
R6141
Ea./
48.14
5000 Units, High Concentration, 40–80 Units/μL Sma I (Xma I) (Blue/White Cloning and Genome Qualified) CCCGGG
GGGCCC
R4124
Ea./
216.00
1000 Units
Sma I (Xma I) (Blue/White Cloning and Genome Qualified) CCCGGG
GGGCCC
R6121
Ea./
78.00
5000 Units
Sma I (Xma I) (Blue/White Cloning and Genome Qualified) CCCGGG
GGGCCC
R6125
Ea./
239.00
100 Units
SnaB I
TACGTA
ATGCAT
R6791
Ea./
59.00
500 Units
SnaB I
TACGTA
ATGCAT
R6795
Ea./
234.00
200 Units
Spe I (Blue/White Cloning and Genome Qualified)
ACTAGT
TGATCA
R6591
Ea./
81.00
1000 Units
Spe I (Blue/White Cloning and Genome Qualified)
ACTAGT
TGATCA
R6595
Ea./
307.00
200 Units
Sph I (Bbu I) (Blue/White Cloning Qualified)
GCATGC
CGTACG
R6261
Ea./
89.00
1000 Units
Sph I (Bbu I) (Blue/White Cloning Qualified)
GCATGC
CGTACG
R6265
Ea./
332.00
continued from previous page
continued on next page
1.800.932.5000
|
vwr.com
|
VWR International
|
325
Molecular Diagnostics
Enzymes
1...,328,329,330,331,332,333,334,335,336,337 339,340,341,342,343,344,345,346,347,348,...955
Powered by FlippingBook