VWR Healthcare Clinical Catalog 2012-2014 - page 337
Size
Enzyme
Recognition/Cut Site
Supplier No. Cat. No.
Price
5000 Units
Hinf I
GANTC
CTNAG
R6205
Ea./
200.00
100 Units
Hpa I
GTTAAC
CAATTG
R6301
Ea./
57.00
500 Units
Hpa I
GTTAAC
CAATTG
R6305
Ea./
210.00
1000 Units
Hpa II
CCGG
GGCC
R6311
Ea./
83.00
5000 Units
Hpa II
CCGG
GGCC
R6315
Ea./
306.00
500 Units
Hsp92 I
G(A/G)CG(T/C)C
C(T/C)GC(A/G)G
R7151
Ea./
57.00
1000 Units
Hsp92 II
CATG
GTAC
R7161
Ea./
73.00
10,000 Units
I-Ppo I (Intron-Encoded Endonuclease) (Genome Qualified) CTCTCTTAAGGTAGC
GAGAGAATTCCATCG
R7031
Ea./
127.00
12,500 Units, High Concentration, 40–80 Units/μL Kpn I (Acc65 I) (Blue/White Cloning Qualified)
GGTACC
CCATGG
R4344
Ea./
190.00
2500 Units
Kpn I (Acc65 I) (Blue/White Cloning Qualified)
GGTACC
CCATGG
R6341
Ea./
70.00
10,000 Units
Kpn I (Acc65 I) (Blue/White Cloning Qualified)
GGTACC
CCATGG
R6345
Ea./
144.00
200 Units
Mbo I (Dpn I, Sau3A I, Nde II)
GATC
CTAG
R6711
Ea./
69.00
100 Units
Mbo II
GAAGA(N)8
CTTCT(N)7
R6723
Ea./
69.00
1000 Units
Mlu I (Blue/White Cloning and Genome Qualified)
ACGCGT
TGCGCA
R6381
Ea./
70.00
10,000 Units, High Concentration, 40–80 Units/μL Msp I
CCGG
GGCC
R4404
Ea./
244.00
2000 Units
Msp I
CCGG
GGCC
R6401
Ea./
70.00
10,000 Units
Msp I
CCGG
GGCC
R6405
Ea./
244.00
1000 Units
MspA1 I
C(A/C)GC(G/T)G
G(T/G)CG(C/A)C
R7021
Ea./
84.00
250 Units
Nae I (NgoM IV)
GCCGGC
CGGCCG
R7131
Ea./
44.00
1000 Units
Nae I (NgoM IV)
GCCGGC
CGGCCG
R7135
Ea./
128.00
250 Units
Nae I (NgoM IV) Turbo
GCCGGC
CGGCCG
R7231
Ea./
49.00
200 Units
Nar I
GGCGCC
CCGCGG
R6861
Ea./
57.00
200 Units
Nar I Turbo
GGCGCC
CCGCGG
R7261
Ea./
66.00
1000 Units
Nci I
CC(C/G)GG
GG(G/C)CC
R7061
Ea./
68.00
200 Units
Nco I (Blue/White Cloning Qualified)
CCATGG
GGTACC
R6513
Ea./
83.00
1000 Units
Nco I (Blue/White Cloning Qualified)
CCATGG
GGTACC
R6515
Ea./
342.00
500 Units
Nde I
CATATG
GTATAC
R6801
Ea./
131.00
200 Units
Nde II (Dpn I, Mbo I, Sau3A I)
GATC
CTAG
R7291
Ea./
49.00
1000 Units
Nde II (Dpn I, Mbo I, Sau3A I)
GATC
CTAG
R7295
Ea./
200.00
250 Units
Nhe I (Genome Qualified)
GCTAGC
CGATCG
R6501
Ea./
74.00
1250 Units
Nhe I (Genome Qualified)
GCTAGC
CGATCG
R6505
Ea./
222.00
1000 Units, High Concentration, 40–80 Units/μL Not I (Blue/White Cloning and Genome Qualified)
GCGGCCGC
CGCCGGCG
R4434
Ea./
281.00
200 Units
Not I (Blue/White Cloning and Genome Qualified)
GCGGCCGC
CGCCGGCG
R6431
Ea./
67.00
1000 Units
Not I (Blue/White Cloning and Genome Qualified)
GCGGCCGC
CGCCGGCG
R6435
Ea./
252.00
200 Units
Nru I
TCGCGA
AGCGCT
R7091
Ea./
54.00
250 Units
Nsi I (Blue/White Cloning Qualified)
ATGCAT
TACGTA
R6531
Ea./
68.00
continued from previous page
continued on next page
324
|
VWR International
|
vwr.com
|
1.800.932.5000
Molecular Diagnostics
Enzymes
1...,327,328,329,330,331,332,333,334,335,336 338,339,340,341,342,343,344,345,346,347,...955
Powered by FlippingBook