VWR Collection Catalogue - page 264
250
Sieving and Filtration
Sieves and Sieve Shakers
Continued from previous page
Description
Pk
Cat. No.
Ultrasonic sieving system to fit 200 mm test sieve, CH-plug
1
Ultrasonic sieving system to fit 200 mm test sieve, EU-plug
1
Ultrasonic sieving system to fit 200 mm test sieve, UK-plug
1
Ultrasonic sieving system to fit 300 mm test sieve, CH-plug
1
Ultrasonic sieving system to fit 300 mm test sieve, EU-plug
1
Ultrasonic sieving system to fit 300 mm test sieve, UK-plug
1
Ultrasonic sieving system to fit 400 mm test sieve, CH-plug
1
Ultrasonic sieving system to fit 400 mm test sieve, EU-plug
1
Ultrasonic sieving system to fit 400 mm test sieve, UK-plug
1
Description
For
Pk
Cat. No.
IQ/OQ documentation
EML 200 digital plus, EML 315 digital plus and EML 450 digital plus sieve
shakers
1
IQ-OQ documentation for VWR-HAVER sieve shakers
VWR by Haver & Boecker
Description
Pk
Cat. No.
HAVER CSA 5.0 BASIC software
1
HAVER CSA 5.0 EXPERT Software
1
HAVER CSA 5.0 NETWORK Software
1
BASIC
EXPERT
NETWORK
Input of sieve sets, materials and customers/
Analysis report /Protocol (DIN 2591 + DIN
66165) /
Particle size account (DIN 66141)
X
X
X
Statistic functions e.g. for materials and
customers
X
X
Expanded account e.g. for Sauter mean
diameter
X
X
Account of AFS-numbers according to VDG-
data sheet
X
X
Definition of particle limits / Loading of
comparable data
X
X
Graphics: RRSB-net, cumulative oversize
distribution curve
X
X
Data bank functions e.g. searching and
reporting
X
X
Data export e.g. to Excel
X
X
Import of aged data
X
X
Take over of external data in the context
of LIM’s systems e.g. numbers of samples
and charges
X
Transfer to external systems (LIMS) by txt-
data connections
X
Evaluation Software for Sieve Analysis, HAVER CSA 5.0
VWR by Haver & Boecker
The HAVER CSA 5.0 software enables PC-aided analysis of standard sieve systems. Three versions are
available which differ in the scale of functions. The results of measurements are documented for quality
assurance purposes and can be introduced into statistics and reports due to the complex data processing.
Ş Simple and precise reports
Ş User friendly and self-explanatory user interface
Ş Complex graphical display and detailed documentation
Ş Operates using systems Windows® XP, 7,0 and Vista systems
Ş Connection to different types of scales and test sieve shakers
1...,254,255,256,257,258,259,260,261,262,263 265,266,267,268,269,270,271,272,273,274,...612
Powered by FlippingBook