VWR Collection Catalogue - page 263
249
Sieving and Filtration
Sieves and Sieve Shakers
Type
Application
Pk
Cat. No.
EML 315 digital plus with ”TwinNut” quick-release clamping system, for sieves up to 315 mm Ø
EML 315 digital plus N with „TwinNut“ quick-release clamping system,
EU-plug
Wet sieving
1
EML 315 digital plus N with „TwinNut“ quick-release clamping system,
UK-plug
Wet sieving
1
EML 315 digital plus N with „TwinNut“ quick-release clamping system,
CH-plug
Wet sieving
1
EML 450 digital plus with ”Classic” clamping system, for sieves up to 450 mm Ø
EML 450 digital plus T, EU-plug
Dry sieving
1
EML 450 digital plus T, UK-plug
Dry sieving
1
EML 450 digital plus T, CH-plug
Dry sieving
1
EML 450 digital plus N, EU-plug
Wet sieving
1
EML 450 digital plus N, UK-plug
Wet sieving
1
EML 450 digital plus N, CH-plug
Wet sieving
1
EML 450 digital plus with ”TwinNut” quick-release clamping system, for sieves up to 450 mm Ø
EML 450 digital plus T with „TwinNut“ quick-release clamping system,
EU-plug
Dry sieving
1
EML 450 digital plus T with „TwinNut“ quick-release clamping system,
UK-plug
Dry sieving
1
EML 450 digital plus T with „TwinNut“ quick-release clamping system,
CH-plug
Dry sieving
1
EML 450 digital plus N with „TwinNut“ quick-release clamping system,
EU-plug
Wet sieving
1
EML 450 digital plus N with „TwinNut“ quick-release clamping system,
UK-plug
Wet sieving
1
EML 450 digital plus N with „TwinNut“ quick-release clamping system,
CH-plug
Wet sieving
1
Description
For
Pk
Cat. No.
Accessories
Reduction piece for cover, for sieve Ø 76 mm
EML 200
1
Reduction piece for cover, for sieve Ø 100 mm
EML 200
1
Reduction piece for cover, for sieve Ø 150 mm
EML 200
1
Reduction piece for cover, for sieve pan with discharge nozzle <400 mm EML 450
1
Inspection glass with central hole
EML 315
1
Inspection glass with central hole
EML 450
1
Continued from previous page
Description
Pk
Cat. No.
Stainless steel sieve pan, 300 mm Ø, 60 mm effective height, for dry sieving
1
Stainless steel sieve pan with discharge nozzle and PVC water hose, 300 mm Ø, 60 mm effective height, for wet sieving
1
Sieve pan and standard sieve set for EML 315 digital plus
VWR by Haver & Boecker
Description
Pk
Cat. No.
Stainless steel sieve pan, 400 mm Ø, 65 mm effective height, for dry sieving
1
Stainless steel sieve pan with discharge nozzle and PVC water hose, 400 mm Ø, 65 mm effective height, for wet sieving
1
Sieve pan and standard sieve set for EML 450 digital plus/UWL 400
VWR by Haver & Boecker
Ultrasonic Sieving System
VWR by Haver & Boecker
Using ultrasonic frequency variation for test sieving, even powders with separating cuts of ≤ 300 micron can
be sieved. Fitting at least one HAVER-Test Sieve with a frequency variation system enables such analyses.
The ultrasonic frequency variation system consists of a clamping ring, which is connected by a converter to
a generator. The generator converts electrical energy into high frequency energy which is then converted to
mechanical energy by the converter and transferred to the clamping ring. The sound waves cause the test
sieve frame to oscillate at high frequency. These oscillations are transferred to the test sieve cloth where
they are evenly distributed.
The oscillations of the test sieve cloth reduce the frictional resistance between the grains and the screen.
This reduces the tendency towards blockage, resulting in increased throughput. The end of a test sieving
period is clearly reached more rapidly. As a result, test sieving of some difficult products is easier and in
some cases now becomes possible..
1...,253,254,255,256,257,258,259,260,261,262 264,265,266,267,268,269,270,271,272,273,...612
Powered by FlippingBook