VWR Collection Catalogue - page 262
248
Sieving and Filtration
Sieves and Sieve Shakers
Type
Application
Pk
Cat. No.
EML 200 digital plus with ”Classic” clamping system, for sieves up to 203 mm Ø
EML 200 digital plus T, EU-plug
Dry sieving
1
EML 200 digital plus T, UK-plug
Dry sieving
1
EML 200 digital plus T, CH-plug
Dry sieving
1
EML 200 digital plus N, EU-plug
Wet sieving
1
EML 200 digital plus N, UK-plug
Wet sieving
1
EML 200 digital plus N, CH-plug
Wet sieving
1
EML 200 digital plus with ”TwinNut” quick-release clamping system, for sieves up to 203 mm Ø
EML 200 digital plus T with „TwinNut“ quick-release clamping system,
EU-plug
Dry sieving
1
EML 200 digital plus T with „TwinNut“ quick-release clamping system,
UK-plug
Dry sieving
1
EML 200 digital plus T with „TwinNut“ quick-release clamping system,
CH-plug
Dry sieving
1
EML 200 digital plus N with „TwinNut“ quick-release clamping system,
EU-plug
Wet sieving
1
EML 200 digital plus N with „TwinNut“ quick-release clamping system,
UK-plug
Wet sieving
1
EML 200 digital plus N with „TwinNut“ quick-release clamping system,
CH-plug
Wet sieving
1
EML 315 digital plus with ”Classic” clamping system, for sieves up to 315 mm Ø
EML 315 digital plus T, EU-plug
Dry sieving
1
EML 315 digital plus T, UK-plug
Dry sieving
1
EML 315 digital plus T, CH-plug
Dry sieving
1
EML 315 digital plus N, EU-plug
Wet sieving
1
EML 315 digital plus N, UK-plug
Wet sieving
1
EML 315 digital plus N, CH-plug
Wet sieving
1
EML 315 digital plus with ”TwinNut” quick-release clamping system, for sieves up to 315 mm Ø
EML 315 digital plus T with „TwinNut“ quick-release clamping system,
EU-plug
Dry sieving
1
EML 315 digital plus T with „TwinNut“ quick-release clamping system,
UK-plug
Dry sieving
1
EML 315 digital plus T with „TwinNut“ quick-release clamping system,
CH-plug
Dry sieving
1
The three electromagnetically driven test sieve shakers are designed for sieve analysis with optimum three-dimensional sieving action. The self re-adjusting
amplitude, which uses permanent acceleration measurement of the whole sieve tower, balances the oscillation of the shaker and its stand. This guarantees that
amplitude stays the same, irrespective of the feed quantity and the installation position.
The EML 200 digital plus T for dry sieving has a built-in control panel. EML 200, 315 and 450 models for wet sieving have a separate control device.
Optional „TwinNut“ quick-release clamping system:
The “TwinNut” clamps for easy fixing and removal of the cover are an alternative to the standard “Classic“ knurled nuts. The guide rods of the “Classic” system can
also be used for the “TwinNut” system.
Ş Memory for 10 test sieve parameter sets
Ş Timer function: 0 - 99 min or continuous operation
Ş Intermittent or continuous shaking operation
Ş Digital display of all functions
Ş 3000 impulses per minute (50 Hertz)
Ş Sound emission: ≤70 dB(A) for EML 200 digital plus and EML 315 digital plus, EML 450 digital plus: 73 dB(A)
Ş Maximum loading: 3 kg
Ordering Information:
IQ-OQ documentation on request.
Delivery Information:
Included as standard with each shaker: Models with either „Classic“ clamping system or „TwinNut“ system are supplied with two 620
mm guide rods and two knurled nuts with handlebars, cover with inspection glass, water spray diffuser for wet sieving models and software for analysis of sieve
results.
Test sieve shakers, EML digital plus
VWR by Haver & Boecker
Continued on next page
1...,252,253,254,255,256,257,258,259,260,261 263,264,265,266,267,268,269,270,271,272,...612
Powered by FlippingBook