VWR Collection Catalogue - page 259
245
Sterilisation and Disinfection
Autoclaves
Description
Pk
Cat. No.
Autoclave, vertical, VAPOUR-Line eco 25, with 5 personal programmes, can hold 1 Ecobox-25 or 2 Ecobox-25-Mini
1
Autoclave, vertical, VAPOUR-Line eco 25, with printer, with 5 personal programmes, can hold 1 Ecobox-25 or 2 Ecobox-25-Mini
1
Autoclave, vertical, VAPOUR-Line eco 50, with 7 personal programmes, can hold 2 Ecobox 50 or 3 Ecobox-50-Mini
1
Description
Pk
Cat. No.
Accessories for Beta Easy Lock Autoclaves
Ecobox 50 basket: (Ø×h): 310×285, in stainless steel AISI 304 (max. loading: 2 baskets)
1
Ecobox-25-Mini: (Ø×h): 240×210, in stainless steel AISI 304 (max. loading: 2 baskets)
1
Ecobox-25 basket: (Ø×h): 240×420, in stainless steel AISI 304 (max. loading: 1 basket)
1
Ecobox-50-Mini basket: (Ø×h): 310×190 in stainless steel AISI 304 (max. loading: 2 baskets)
1
Data printer
1
Model
VAPOUR-Line eco 25
VAPOUR-Line eco 50
Voltage (V)
220
380 (triphase)
Weight (kg)
45
60
W×D×H ext. (mm)
550×540×1005
550×570×1005
Continued from previous page
Vertical autoclaves, VAPOUR-line
The housing is made from powder coated steel
VAPOUR-Line autoclaves are ideal for different laboratory applications, such as: Liquids in open or slightly closed
vessels, instruments and solids, unwrapped and waste sterilisation.
All VAPOUR-Line 80 and 135 models generate steam directly in the chamber. Heating elements are situated
under the base of the chamber so there is no contact between them and feed water, ensuring long element
lifetime and preventing contamination of the elements by spilt waste, broken bottles or similar. The elements are
protected from overheating by a temperature limiter.
VAPOUR-Line 80M and 135M models are equipped with an effective air cooling system for liquid sterilisation.
This speeds cooling time by up to 40% compared to standard models.
Ş Equiped as standard with a flexible PT100 temperature sensor
Ş Equiped with a microprocessor controller with 5 pre-programmed programmes
Ş TÜV approved safety locking mechanism and thermal lock according EN DIN 61010-2-43
Ş Air removal by gravity
Ş Integrated exhaust steam condensation, water cooled and temperature controlled
Ş Integrated memory chip for storing up to 150 cycles (later printout possible)
Ş RS 485 interface
Description
Pk
Cat. No.
Vertical autoclave, VAPOUR-Line 80, EU-plug
1
Vertical autoclave, VAPOUR-Line 80, UK-plug
1
Vertical autoclave, VAPOUR-Line 135, EU-plug
1
Vertical autoclave, VAPOUR-Line 135, UK-plug
1
Model
VAPOUR- line 80/80M
VAPOUR- line 135/135M
Capacity (l)
80
135
Heating power (W)
300
600
Internal Ø×H (mm)
410×600
500×660
Temperature (°C)
98...138
Voltage (V)
230
Weight (kg)
135/145
160/170
W×D×H ext. (mm)
740×915×600
840×965×700
Basic units
Description
Pk
Cat. No.
Vertical autoclave, VAPOUR-Line 80 M, EU-plug, with integrated cooling system for liquid sterilisation
1
Vertical autoclave, VAPOUR-Line 80 M, UK-plug, with integrated cooling system for liquid sterilisation
1
Vertical autoclave, VAPOUR-Line 135 M, EU-plug, with integrated cooling system for liquid sterilisation
1
Vertical autoclave, VAPOUR-Line 135 M, UK-plug, with integrated cooling system for liquid sterilisation
1
Model
VAPOUR- line 80/80M
VAPOUR- line 135/135M
Capacity (l)
80
135
Heating power (W)
300
600
Internal Ø×H (mm)
410×600
500×660
Temperature (°C)
98...138
Voltage (V)
230
Weight (kg)
135/145
160/170
W×D×H ext. (mm)
740×915×600
840×965×700
Air cooled units
Accessories
Continued on next page
1...,249,250,251,252,253,254,255,256,257,258 260,261,262,263,264,265,266,267,268,269,...612
Powered by FlippingBook