VWR Collection Catalogue - page 258
244
Sterilisation and Disinfection
Autoclaves
Description
Pk
Cat. No.
Bench top vertical autoclave, VWR VAPOUR-Line lite
1
Bench top vertical autoclave, with integrated printer, VWR VAPOUR-Line lite P
1
Description
Pk
Cat. No.
Accessories
LITEbox basket: (Ø×h): 280×260, with transporting handle, in stainless steel
1
Model
VWR VAPOUR-Line lite
VWR VAPOUR-Line lite P
Capacity (l)
23
External Ø×H (mm)
470×580
Internal Ø×H (mm)
310×290
Material
exterior: stainless steel AISI 304
interior: aluminium
Power rating (kW)
1300
Temperature (°C)
121 (max)
Weight (kg)
13,250
Autoclave, VWR VAPOUR-Line lite
AISI 340 stainless steel housing, easy to maintain and disinfect, aluminium inner chamber and ABS insula-
ting lid with locking system.
Ş Microprocessor controlled automatic thermoregulation
Ş Digital temperature and sterilisation time display
Ş Automatic ventilation valve and electronic temperature cut-off safety protection system
Ş Reinforced heating element and PT100 accurate thermoprobe
Ş Automatic sterilisation programmes
Instructions are GLP and EN 17205 compliant.
Ordering Information:
Basket has to be ordered separately.
Model
VAPOUR-Line eco 25
VAPOUR-Line eco 50
Capacity (l)
25
50
Internal Ø×H (mm)
250×450
320×600
Material
Stainless steel 304
Max. pressure (bar)
2,1
Power rating (kW)
1,7
3,7
Temperature (°C)
134
Inside pressure vessel and outside structure made in AISI 304 stainless steel mirror finished grade
Easy to use, automatic, flexible, reliable and convenient autoclave for all standard laboratory procedures with some special features as F0 calculation and liquid
sterilisation with flexible thermoprobe (for 50 l model).
Ş User friendly programmer, instructions printed on the panel
Ş Easy to install, easy to use, easy to maintain
Ş Fully automatic with GLP special functions
Ş Reliable: 10 years warranty against perforating corrosion
Ş Customisable programmes
Ş Safety: ABS insulating lid with vertical position lock – temperature dependent lid lock
Ş Flexible PT100 thermoprobe for eco 50 (fixed for eco 25), can be positioned into the liquid load (max 1 litre recommended)
Conform to European PED regulations, CE mark. IQ, OQ, PQ documentation available on request.
Top Loading VAPOUR-Line Eco autoclaves
Continued on next page
1...,248,249,250,251,252,253,254,255,256,257 259,260,261,262,263,264,265,266,267,268,...612
Powered by FlippingBook