VWR Collection Catalogue - page 257
243
Cold Treatment
Cryo Storage Systems
Description
Dimensions (mm)
Pk
Cat. No.
Labels, blue strips for 1,5 - 2,0 ml tubes
33×13
1.000
Labels, green strips for 1,5 - 2,0 ml tubes
33×13
1.000
Labels, multi-coloured strips for 1,5 ml
tubes, suitable for laser printers
24×13
2.125
Labels, multi-coloured strips for 96/384 well
plates, suitable for laser printers
38×6
3.900
Labels, purple strips for 1,5 - 2,0 ml tubes 33×13
1.000
Labels, red strips for 1,5 - 2,0 ml tubes
33×13
1.000
Labels, white strips for 0,2 ml PCR tubes
20×6
1.000
Labels, white strips for 0,5 ml tubes, suitab-
le for laser printers
24×13
2.380
Labels, white strips for 1,5 - 2,0 ml tubes
33×13
1.000
Labels, white strips for 1,5 ml tubes, suitab-
le for laser printers
33×13
1.700
Labels, white strips for 96/384 well plates,
suitable for laser printers
38×6
3.120
Labels, white strips for racks and boxes,
suitable for laser printers
67×25
600
Labels, yellow strips for 1,5 - 2,0 ml tubes 33×13
1.000
Labels, square
The labels and adhesive have been designed to withstand temperatures from –196 to +121 °C. VWR labels
are ideal for all cryostorage and can also be used in most incubators.
Ş No need to laminate
Ş Use on tubes, racks and boxes
1...,247,248,249,250,251,252,253,254,255,256 258,259,260,261,262,263,264,265,266,267,...612
Powered by FlippingBook