VWR Collection Catalogue - page 256
242
Cold Treatment
Cryo Storage Systems
Continued from previous page
W×D×H ext. (mm)
Compartments
For (ml)
Colour
Pk
Cat. No.
133×133×52
10×10
cryotubes 1 - 2
Yellow
4
133×133×81
9×9
cryotubes 3 - 4
Blue
3
133×133×81
9×9
cryotubes 3 - 4
Green
3
133×133×81
9×9
cryotubes 3 - 4
Red
3
133×133×81
9×9
cryotubes 3 - 4
Yellow
3
W×D×H ext. (mm)
Compartments
Type
Pk
Cat. No.
Liquid nitrogen cryogenic freezer boxes with drain slots
70×70×47,6
25
-
1
127×125×47,6
100
-
1
Mechanical cryogenic freezer boxes with or without dividers
127×127×47,6
81
With dividers
1
127×127×51
-
Without dividers
1
127×127×73
-
Without dividers
1
Fiberboard dividers
125×125×25,4
16
-
12
125×125×25,4
25
-
12
125×125×25,4
49
-
1
125×125×25,4
49
-
12
125×125×25,4
64
-
1
125×125×25,4
64
-
12
125×125×25,4
81
-
1
125×125×25,4
81
-
12
125×125×25,4
100
-
1
125×125×25,4
100
-
12
Cryostorage boxes and dividers
Boxes and dividers are constructed from durable fibreboard with a protective, moisture-repellant coating.
Liquid nitrogen (LN
2
) freezer boxes feature slots for safe LN
2
drainage and reduced LN
2
consumption.
Mechanical freezer boxes do not feature slots and cannot be used with LN
2
.
Boxes with dividers feature a lid printed with a numbered grid for superior inventory control. Boxes without
dividers can accommodate dividers (sold separately) for 16 to 100 places. Fibreboard dividers can fit in any
127x127 mm LN
2
cryogenic or mechanical freezer box.
Compartments
W×D×H ext. (mm)
Cryobox height (mm)
Pk
Cat. No.
12 (3×4)
556×140×245
75
1
16 (4×4)
556×140×225
50
1
Racks for cryoboxes
Stainless steel, with handles
Horizontal racks fit cryoboxes up to 133×133 mm.
Compartments
W×D×H ext. (mm)
Pk
Cat. No.
Racks without cryoboxes
16 (4×4)
562×139×225
1
20 (5×4)
562×139×285
1
24 (6×4)
562×139×331
1
Racks with standard white cardboard cryoboxes with 9×9 cell dividers
16 (4×4)
562×139×225
1
20 (5×4)
562×139×285
1
24 (6×4)
562×139×331
1
Racks for cryoboxes
Cardboard
Economical coated cardboard racks for upright freezers.
Ş Stable and long lasting
Ş Lightweight - the 20 compartment rack weighs 1,1 kg without boxes
Ş For cryoboxes 50 mm height
1...,246,247,248,249,250,251,252,253,254,255 257,258,259,260,261,262,263,264,265,266,...612
Powered by FlippingBook