VWR Collection Catalogue - page 255
241
Cold Treatment
Cryo Storage Systems
Description
No. of holes
Pk
Cat. No.
−20 °C mini labtop cooler
12
1
−20 °C maxi labtop cooler
32
1
−20 °C labtop cooler
20
1
Labtop coolers
−20 °C mini labtop cooler PC/non toxic gel
The −20 °C mini labtop cooler holds 12×0,5 or 1,5 ml microcentrifuge tubes at below −20 °C for over 1
hour. Ideal for bench top use or transport. Wire handle secures top while carrying. Filled with non toxic gel.
Prior to use, place in −25 °C freezer for at least 24 hours, store at −25 °C between use.
−20 °C maxi labtop cooler PC/non toxic gel
The −20 °C maxi labtop cooler, holds 32×0,5 or 1,5 ml microcentrifuge tubes at below −20 °C for over 1
hour. Suitable for bench top use or transport. Wire handle secures top while carrying. Cooler and lid filled
with non toxic gel. Store at −25 °C between use.
−20 °C labtop cooler PC/non toxic gel
The −20 °C labtop cooler maintains temperatures between −20 and −15 °C for up to four hours. Polycar-
bonate. Holds twenty 1,5 to 2,0 ml microcentrifuge tubes or cryovials. Inserts included hold 0,2 to 0,5 ml
microcentrifuge tubes. Printed grid, keyed lid, locking handle, non skid feet. Store at −25 °C between use.
Description
I-Ø×H (mm)
No. of holes
Pk
Cat. No.
Cryo 1 °C freezing container 86×117
18
1
Cryo 1° C freezing container
Container PC, closure and vial holder HDPE, foam insert
Non mechanical device that uses isopropyl alcohol and a mechanical freezer to provide cooling rate of –1 °C
per minute. Ideal for successful cell cryopreservation and recovery.
Ş Floating rigid insert prevents vials from contamination
Ş Withstands repeated use; Labelled with step by step instructions
Ş Holds up to 18 cryogenic vials (1; 1,2; 1,5 and 2 ml)
W×D×H ext. (mm)
Compartments
For (ml)
Colour
Pk
Cat. No.
76×76×52
5×5
cryotubes 1 - 2
Blue
8
76×76×52
5×5
cryotubes 1 - 2
Green
8
76×76×52
5×5
cryotubes 1 - 2
Red
8
76×76×52
5×5
cryotubes 1 - 2
Yellow
8
133×133×52
9×9
cryotubes 1 - 2
Blue
4
133×133×52
9×9
cryotubes 1 - 2
Green
4
133×133×52
9×9
cryotubes 1 - 2
Red
4
133×133×52
9×9
cryotubes 1 - 2
Yellow
4
133×133×95
9×9
cryotubes 3 - 5
Blue
5
133×133×95
9×9
cryotubes 3 - 5
Green
5
133×133×95
9×9
cryotubes 3 - 5
Red
5
133×133×95
9×9
cryotubes 3 - 5
Yellow
5
133×133×52
10×10
cryotubes 1 - 2
Blue
4
133×133×52
10×10
cryotubes 1 - 2
Green
4
133×133×52
10×10
cryotubes 1 - 2
Red
4
Cryoboxes
PC, autoclavable
Can be used from –190 to +121 °C.
Ş Transparent cover with numeric grid and key to prevent misalignment
Ş Coloured grid or base
1...,245,246,247,248,249,250,251,252,253,254 256,257,258,259,260,261,262,263,264,265,...612
Powered by FlippingBook