VWR Collection Catalogue - page 254
240
Cold Treatment
Cryo Storage Systems
Colour
Pk
Cat. No.
Amber
500
Blue
500
Green
500
Grey
500
Lilac
500
Orange
500
Pink
500
Red
500
Violet
500
White
500
Yellow
500
Set
500
Coloured Capinsert™ for cryogenic vials
PP
Inserts fit precisely into the cap of the cryogenic vial for colour identification.
Set; assortment of white, blue, red, green and yellow (100 of each)
Length (mm)
Pk
Cat. No.
275
100
Sleeves for CryoCanes®
PVC
Protects cryocanes during handling and storage.
For (ml)
No. of holes
W×D×H (mm)
Pk
Cat. No.
(Ø 12,5 mm) 1 - 5
50 (5×10)
199×104×24,5
1
Racks for cryogenic vials
PC, autoclavable
Withstand both cryogenic freezing and autoclaving.
Ş With numeric index of the holes
Ş Moulded tabs in the bottom of each well to secure vials in place for easier opening and closing
Ş Ribbed grips make handling with gloves easier
For (ml)
No. of holes
W×D×H (mm)
Pk
Cat. No.
1 - 5
50 (5×10)
205×103×26
4
Workstation racks for cryogenic vials
PP
A universal locking system locks the vials in each well, so you can easily unscrew them with one hand.
Ş Alphanumeric identification of each well
Ş Strong handles make it easy and safe to carry
Ş Supported by five anti-skid rubber feet, stackable
1...,244,245,246,247,248,249,250,251,252,253 255,256,257,258,259,260,261,262,263,264,...612
Powered by FlippingBook