VWR Collection Catalogue - page 253
239
Cold Treatment
Cryogenic Technique
Description
Pk
Cat. No.
Accessories
Stainless steel rack to be used with cardboard boxes for blood bags, for 8 boxes (horizontally positioned) height: 688 mm
1
Pie-shaped rack system, made of high quality polycarbonate, withstands extremely low temperatures, with 6 racks, each with 9 drawers, 95 vials per
drawer (total 5130 vials)
6
Plastic canisters, (Ø×H: 70×660 mm); each canister can hold 5 goblets (not included), with goblet lifter
1
Stainless steel rack for 10 boxes, to be used with standard cryo-boxes (not included), total height 655 mm
1
Stainless steel rack for 12 boxes, to be used with standard cryo-boxes (not included), total height 655 mm
1
Stainless steel rack for 13 boxes, to be used with standard cryo-boxes (not included), total height 708 mm
1
Stainless steel rack to be used with cardboard boxes for blood bags; rack for 6 boxes for blood bags, height 586 mm
1
Rack system for 2 ml vials, with 6 aluminium pie-shaped racks, each with 13 drawers, 130 vials per drawer (total: 10140), for AF-110
1
Rack system with 3 × 2 aluminium triangular racks, each with 13 drawers and 130 vials per drawer (total 10140 vials), for AF-110
1
Plastic canisters, to be used with goblets Ø 70 mm × H 515 mm for 4 goblets, with goblet lifter
1
Stainless steel canisters, to be used with goblets: Ø 70 mm × H 525 mm for 4 goblets, with goblet lifter
1
Stainless steel canister, (Ø×H: 70×660 mm), each canister can hold 5 goblets (not included), with goblet lifter
1
Phase separator, Ø×H: 30×100 mm
1
Cardboard box, each for 2 blood bags DF700
1
Cassette for DF700 blood bag
1
Hot-gas bypass
1
Battery back-up (24 hours)
1
Standard flexible hose; 1,20 m, 2×female ¾“ connections
1
Standard flexible hose; 2 m, 2×female ¾“ connections
1
Standard flexible hose; 3 m, 2×female ¾“ connections
1
Optional software to monitor all functions of the S-170 controller on PC (including 1 cable for 1 controller)
1
Optional software to monitor all functions of the S-170 controller on PC (including 1 cable for 1 controller)
1
Continued from previous page
Capacity (ml)
Base
Ø ext.×H (mm)
Pk
Cat. No.
With external thread and lip seal
1,2
Self-standing
12,5×41
250
2,0
Round bottom
12,5×46
250
2,0
Self-standing
12,5×48
250
5,0
Self-standing
12,5×90
250
With internal thread and silicone O-ring
1,2
Self-standing
12,5×41
250
2,0
Round bottom
12,5×48
250
2,0
Self-standing
12,5×49
250
4,0
Round bottom
12,5×70
250
4,0
Self-standing
12,5×72
250
5,0
Round bottom
12,5×90
250
Cryovials with lip seal and silicone washer - external threads
1,2
Self-standing
12,5×43
250
2,0
Round bottom
12,5×48
250
2,0
Self-standing
12,5×49
250
3,0
Self-standing
12,5×72
250
4,0
Self-standing
12,5×76
250
5,0
Self-standing
12,5×93
250
Cryovials with silicone washer - internal threads
1,2
Self-standing
12,5×41
250
2,0
Round bottom
12,5×48
250
2,0
Self-standing
12,5×49
250
4,0
Round bottom
12,5×70
250
4,0
Self-standing
12,5×72
250
5,0
Round bottom
12,5×90
250
5,0
Self-standing
12,5×92
250
With external thread and silicone O-ring
1,2
Self-standing
12,5×43
250
2,0
Round bottom
12,5×48
250
2,0
Self-standing
12,5×49
250
3,0
Self-standing
12,5×72
250
4,0
Self-standing
12,5×76
250
5,0
Self-standing
12,5×93
250
10,0
Self-standing
17,0×84
250
Cryogenic vials
PP vial and closure, sterile
Designed for the storage of biological material, human or animal cells, at temperatures as low as -196 °C
(but should be used only in the gas phase of liquid nitrogen).
Ş Certified RNase-, DNase-, pyrogen- and DNA- free
Ş With graduation and marking area
Round bottom vials can be centrifuged up to 17 000×g
1...,243,244,245,246,247,248,249,250,251,252 254,255,256,257,258,259,260,261,262,263,...612
Powered by FlippingBook