VWR Collection Catalogue - page 252
238
Cold Treatment
Cryogenic Technique
Type
Pk
Cat. No.
LN2 cylinder, CryoCyl-60
1
LN2 cylinder, CryoCyl-120
1
LN2 cylinder, CryoCyl-230
1
Description
Pk
Cat. No.
Accessories for CryoCyl
Level gauge for CryoCyl-60
1
Level gauge for CryoCyl-120
1
Level gauge for CryoCyl-230
1
Model
CryoCyl-60
CryoCyl-120
CryoCyl-230
Capacity (l)
58
120
230
Weight, empty (kg)
38
92
133
W×D×H ext. (mm)
-
685×725×1397
Ø×H (mm)
460×1000 (without gauge)
570×1205
-
Liquid nitrogen cylinders, Cryocyl
Type
Pk
Cat. No.
Liquid storage container, AF-75
1
Vapour storage container, AF-75P
1
Dry storage in square racks, AF-75DS
1
Liquid storage container, AF-110 (EU plug version)
1
Liquid storage container, AF-110 (UK plug version)
1
Liquid storage container, AF-110 (CH plug version)
1
Liquid storage container, AF-200
1
Vapour storage container, AF-200P
1
Dry storage in square racks, AF-200DS
1
Liquid storage container, AF-250
1
Vapour storage container, AF-250P
1
Dry storage in square racks, AF-250DS
1
Liquid storage container, AF-400 (EU plug version)
1
Liquid storage container, AF-400 (UK plug version)
1
Liquid storage container, AF-400 (CH plug version)
1
Vapour storage container, AF-400P (EU plug version), with rotating platform
1
Vapour storage container, AF-400P (UK plug version), with rotating platform
1
Vapour storage container, AF-800P (EU plug version), with rotating platform and narrow neck opening (420 mm)
1
Vapour storage container, AF-800P (UK plug version), with rotating platform and narrow neck opening (420 mm)
1
Vapour storage container, AF-1200P (EU plug version), with rotating platform and narrow neck opening (520 mm)
1
Vapour storage container, AF-1200P (UK plug version), with rotating platform and narrow neck opening (520 mm)
1
Model
AF-75 AF-75P
AF-
75DS
AF-110 AF-200
AF-
200P
AF-
200DS
AF-250
AF-
250P
AF-
250DS
AF-400
AF-
400P
AF-800P
AF-
1200P
Capacity
(l)
75
110
198
236
401
790
1240
Weight,
empty
(kg)
37
80
70
72
300
320
315
460
Liquid nitrogen auto-fill system, AF series
The AF series units are large capacity, auto-fill systems offering a fully-featured controller with an easy to
use interface. The control module is designed to manage the cryogenic tank automatically. The units need
LN2 supply from a stand-alone pressurised vessel, or from a central LN2 supply system.
The
AF-75
,
-200
and
-250
have an aluminium inner and outer vessel. The units can be mounted on optio-
nal castors. The
S-170
controller has a separate housing attached to the side of the vessels. The lids can be
locked with a pad lock (not included, not possible on
AF-75
).
The
AF-110
has a round aluminium inner vessel and a square outer casing of powder coated metal. The
unit is mounted on castors. The
AF-400
has a stainless steel inner vessel. The user can choose a rotating
platform on this model (
AF-400P
). This facilitates access to the racks in the back, and reduces the number
of racks. Castors and lock are standard mounted. On both models the S-170 controller is built-into the front
panel for easy setting and reading.
The
AF-800P
and
AF-1200P
full stainless steel biological containers are designed for storage of samples in
the most efficient and safe conditions in liquid or gaseous phase. Consumption of liquid nitrogen is reduced
and holding time is increased thanks to the narrow neck diameter. The valuable samples will be stored in gaseous phase even at -150 °C right under the lid. The
samples are easily accessible thanks to an inner rotating tray, which can be turned around by means of a turning handle located on the top of the container.
The newly developed
S-170 controller
is mounted in a separate housing on the top of the vessel and offers maximum sample security and many features:
automatic filling procedure, temperature measurement (via 2 probes), regulation of liquid nitrogen level, providing visual and audible alarms (low level alarm,
low level fill, normal level stop fill, high level alarm), display of the position of the cover (open/closed), management of the „defog“ function (for better visibility
inside the vessel) and 4 potential-free contacts for connection to an in-house alarm system or to a telephone dialing system (4 different alarm functions can be
transferred). The RS485 interface enables complete management and control of the system via a PC (the software is included).
Continued on next page
1...,242,243,244,245,246,247,248,249,250,251 253,254,255,256,257,258,259,260,261,262,...612
Powered by FlippingBook