VWR Collection Catalogue - page 251
237
Cold Treatment
Cryogenic Technique
Type
Neck-Ø (mm)
Pk
Cat. No.
Dry shipper, DS-2
35
1
Dry shipper, DS-2 including datalogger (2 years average duration)
35
1
Dry shipper, DS-5
70
1
Dry shipper, DS-5 including datalogger (2 years average duration)
70
1
Dry shipper, DS-24
215
1
Dry shipper, DS-24 including datalogger (2 years average duration)
215
1
Description
For
Pk
Cat. No.
Removable cartridges for DS-24
60 mm cartridge to enlarge the capacity of absorbed LN2
DS-24
1
120 mm cartridge to enlarge the capacity of absorbed LN2
DS-24
1
Extractor for cartridges
DS-24
1
Protective flight case for DS, with foam protection inside
Case with 4 mm foam protection inside
DS-2
1
Case with 4 mm foam protection inside
DS-5
1
Case with 7 mm foam protection inside
DS-24
1
Accessories
Phase separator, Ø×H: 30×100 mm
1
Optional rack with 6 drawers, stainless steel for DS-24 without cartridge
1
Optional rack with 5 drawers, stainless steel for DS-24 with 60 mm
cartridge
1
Optional rack with 4 drawers, stainless steel for DS-24 with 120 mm
cartridge
1
Optional rack with 3 drawers, stainless steel for DS-24 with 60+120 mm
cartridge
1
Standard flexible hose; 1,20 m, 2×female ¾“ connections
1
Standard flexible hose; 2 m, 2×female ¾“ connections
1
Standard flexible hose; 3 m, 2×female ¾“ connections
1
RECWIN software: CD-ROM with cables compatible with Windows 95, 98,
2000 NT and XP. Graphic user interface, multi-language (UK, FR, ES, D, I)
1
Replacement battery (recommended every 2 years)
1
Model
DS-2
DS-5
DS-24
Capacity (l)
2
5,2
24
No. of canisters
1
7*
No. of test tube racks
-
1×6 boxes*
Static holding time
(days)
23
19
11
Weight, empty (kg)
3,3
6,9
15,6
Ø×H (mm)
190×407
251×497
400×664
* DS-24 is delivered without canister, rack or cartridge (must be ordered separately). For more details please contact VWR.
Dry shippers, DS-series
Dry shippers are used for the safe transportation of samples at cryogenic temperatures.
Hydrophobic absorbent material is fixed in the inner capacity of the vessel. After complete saturation of the
material with liquid nitrogen, the vessels are ready to transport samples at cryogenic temperatures, without
the risk of liquid nitrogen spilling, if the container is overturned.
Ş Safe transportion of samples
The model DS-24 can be equipped with removable absorbent cartridges to improve the holding time.
The shippers do not conform to International Air Transport Regulations (IATA) for transportation of infec-
tious material.
Certification: CE - Medical
1...,241,242,243,244,245,246,247,248,249,250 252,253,254,255,256,257,258,259,260,261,...612
Powered by FlippingBook