VWR Collection Catalogue - page 248
234
Cold Treatment
Refrigerators and Freezers
Continued from previous page
Type
Pk
Cat. No.
ULT upright freezer, NOVAfreezer, CDW-86L388
1
ULT upright freezer, NOVAfreezer with double solid outer doors, CDW-86L490
1
ULT upright freezer, NOVAfreezer, CDW-86L628
1
ULT upright freezer, NOVAfreezer, CDW-86L728
1
Model
DW-86L388
DW-86L490
DW-86L628
DW-86L728
Display
LED
Door lock
Yes
Door(s)
4 inner/ outer
4 inner/2 outer
4 inner/ outer
Material
Steel
Shelves/drawers
3
Temperature (°C)
− 50...− 86
− 40...− 86
Weight (kg)
254
295
301
350
W×D×H ext. (mm)
890×870×1970
860×900×1970
1035×900×1960
1145×900×1970
W×D×H int. (mm)
620×540×1300
590×630×1310
760×620×1300
870×630×1310
Type
Neck-Ø (mm)
Pk
Cat. No.
CSS series; models with very low LN2 consumption
CSS-4/6
51
1
CSS-10/6
51
1
CSS-21/6
51
1
CSS-36/6
51
1
XSS series; models with low LN2 consumption and increased storage capacity
XSS-13/6
89
1
XSS-26/10
89
1
XSS-36/6
89
1
XSS-48/10
119
1
Description
Pk
Cat. No.
Accessories
Roller base with 4 castors (max. load on a smooth floor: 60 kg)
1
Roller base with 5 castors (max. load on a smooth floor: 250 kg)
1
Roller base with 5 castors (max. load on a smooth floor: 120 kg)
1
Model
CSS-4/6
CSS-10/6
CSS-21/6
CSS-36/6
XSS-13/6
XSS-26/10
XSS/36/6
XSS-48/10
Capacity (l)
4,1
10,5
21,7
35,9
13
26
35,9
48,5
No. of canisters
6
10
6
10
Static holding time
(days)
42 (without
canister)
93 (without
canister)
238 (without
canister)
365 (without
canister)
59 (without
canister)
104 (without
canister)
211 (without
canister)
180 (without
canister)
Weight, empty
(kg)
4,2
6,4
9,8
16
8,7
11,2
14,7
17,1
Ø×H (mm)
245×433
368×455
395×673
480×652
380×450
480×482
480×688
500×707
Ø×H canisters
(mm)
37×110
110×37
37×270
66×110
66×270
73×270
Cryogenic storage vessels, CSS and XSS series
The CSS series tanks offer storage for small to medium capacity applications. The XSS series offer similar
benefits and have extended storage capacity.
The tanks, manufactured from aluminium alloy with fibreglass/epoxy neck. High vacuum, multilayered
super insulation limits nitrogen evaporation.
The samples are placed in stainless steel canisters directly in the liquid nitrogen inside the vessel.
Ş Extremely low liquid nitrogen evaporation
Ş Lid is lockable with padlock (not included)
Ş The vessels are equipped with stainless steel canisters with hook for easy retrieval from the vessel
Canisters which are 270 mm high can hold two goblets or canes with 6×1,8 ml vials
Canisters which are 110 mm high can hold one goblet or 1/2 height canes with 3×1,8 ml vials
Certification: CE - Medical
1...,238,239,240,241,242,243,244,245,246,247 249,250,251,252,253,254,255,256,257,258,...612
Powered by FlippingBook