VWR Collection Catalogue - page 247
233
Cold Treatment
Refrigerators and Freezers
Type
Pk
Cat. No.
Three shelves
1
Four shelves
1
Five shelves
1
Five shelves
1
Model
Capacity (l)
180
260
360
500
Defrost
+
Door lock
Door(s)
One interchangeable door
Specifications
ABS tank
Temperature (°C)
+2...10
Weight (kg)
36
47
62
78
W×D×H ext. (mm)
602×600×884
602×600×1215
602×600×1589
755×715×1515
Temperature: +2 to 10 °C
Refrigerators with explosion-proof interiors, for flammable materials. Ignition sources such as light switches or temperature controllers, which could cause flam-
mable materials to ignite, must not be located in the interior.
Interior lining made from white, high impact special plastic. Smooth surfaces are easy to clean.
Ş Temperature controller attached to the outside
Ş Lockable door
Ş Automatic defrost-function with drip tray for condensation water (manual drainage)
Ş Depending on the model, three to five height adjustable glass shelves for flexible partitioning of the interior space
Ş Adjustable front feet for levelling and firm footing
Refrigerators, explosion-proof
Model
DW-86L388
DW-86L490
DW-86L628
DW-86L728
Air circulation
Ventilated
Alarm
Audible and visible: high and low internal temperature, alarm power failure
Capacity (l)
388
490
628
728
Defrost
Automatic
Upright, ultra-low temperature freezers, NOVAfreezer
Temperature range:−40 to −86 °C
These freezers are specially designed and manufactured for long-term storage of various products, inclu-
ding viruses, germs, leucocytes and cutis. Applications can be found in blood banks, hospitals, epidemic
prevention services, research institutes, biological engineering institutes and marine fishery companies.
The frame is made of steel for maximum durability, insulating walls VIP limited thickness (cavity with poly-
urethane foam panels and vacuum) for maximum insulation of the structure and an optimum ratio between
internal capacity and external dimensions.
Every freezer has a controller with built in software, on the front door, which shows all the necessary compo-
nents (compressors, voltage, temperature sensor) to help quickly identify the cause of a fault. This is called
System monitoring and reporting technology and can be easily accessed through the front panel.
Ş Cascade refrigeration system using two different CFC-free refrigerants (chlorofluorocarbons) and HCFCs
(hydro chlorofluorocarbons)
Ş Solid door handle can be locked with a padlock for maximum security of the contents
Ş With retractable wheels to facilitate movement
Ş With heated triple gasket that provides maximum sealing between the door and swing mechanism
Ş Intelligent condenser design to save energy
Continued on next page
1...,237,238,239,240,241,242,243,244,245,246 248,249,250,251,252,253,254,255,256,257,...612
Powered by FlippingBook