VWR Collection Catalogue - page 246
232
Temperature Treatment and Drying
Desiccators
Type
Ø (mm)
Pk
Cat. No.
Porcelain
90
1
Porcelain
140
1
Porcelain
190
1
Porcelain
235
1
Porcelain
280
1
Desiccator plates
Porcelain desiccator plates. Central hole 20 mm Ø, filter holes 5 mm Ø. Suitable for use with any desicca-
tors with corresponding diameter.
Conform to DIN 12911
1...,236,237,238,239,240,241,242,243,244,245 247,248,249,250,251,252,253,254,255,256,...612
Powered by FlippingBook