VWR Collection Catalogue - page 244
230
Temperature Treatment and Drying
Drying Ovens and Heating Chambers
Description
Pk
Cat. No.
VENTI-Line 53
1
VENTI-Line 115
1
Description
For
Pk
Cat. No.
Shelves
Chrome plated shelf
DL 53/VL 53/IL 53
1
Chrome plated shelf
DL 115/VL 115/IL 115
1
Model
VL 53
VL 115
Capacity (l)
53
115
Convection type
Forced convection
External W×D×H (mm)
620×640×680
820×710×760
Internal W×D×H (mm)
401×330×401
600×400×480
Max. load per shelf (kg)
10
15
Nominal power consumption (W)
800
1000
Shelves supplied (max.)
2 (4)
Temperature fluctuation (time) (°C)
At 150 °C: ±0,4
At 150 °C: ±0,3
Temperature range (°C)
Ambient +12...220
Ambient +5...220
Temperature variation (spatial) (°C)
At 150 °C: ±2,7
At 150 °C: ±2,5
Temperature variation at 150 °C (°C)
±0,4
±0,3
Weight (kg)
40
59
Ovens with forced convection, VENTI-Line®
Drying ovens with forced air circulation, which are ideal for routine drying and heating applications when short
drying times are required. Ovens feature adjustable ventilation by means of manually adjustable ventilation
slide and 50 mm Ø exhaust duct at the rear of the unit.
ŞMicroprocessor controller
Ş Integrated timer can be set for continuous mode or timer operation: 0 to 999 min or 0 to 99,9 h
ŞOver-temperature protection with visual alarm (Class 3,1 safety device)
ŞDigital temperature setting with degree accuracy
Delivery Information:
Supplied with two chrome plated shelves.
Type
Nominal size (DN)
Ø ext.×H (mm)
Ø int. (mm)
Pk
Cat. No.
With stopcock
100
153×180
92
1
With stopcock
150
215×250
143
1
With stopcock
200
207×300
192
1
With stopcock
250
320×355
239
1
With stopcock
300
380×435
258
1
Vaccuum Desiccators
Borosilicate glass 3.3
ŞTubulated lid
ŞStopcock with PTFE spindle
ŞDIN 12491
1...,234,235,236,237,238,239,240,241,242,243 245,246,247,248,249,250,251,252,253,254,...612
Powered by FlippingBook