VWR Collection Catalogue - page 243
229
Temperature Treatment and Drying
Water Baths, Oil Baths and Shaking Water Baths
Description
Pk
Cat. No.
Water bath protective agent, Aquaresist
100 ml
Water bath protective agent, Aquaresist
Prevents the growth of algae and bacteria in baths and circulating thermostats, for optimally hygienic
working conditions.
Ş Durable. Effective for weeks, as displayed by a colour indicator
Ş Economical. Low consumption - only 1 ml solution per 1 litre of water
Description
Pk
Cat. No.
Anti-evaporation spheres
100
Anti-evaporation spheres
1.000
Anti-evaporation spheres for water baths
Lightweight, economic PP spheres which form a blanket on the liquid surface. Spheres have 20 mm Ø, 275
spheres cover approximately 1000 cm
2
.
Ş Reduce heat loss by 77%, and evaporation by 87%
Ş Insulate like a solid, and still allow instant access to the liquid in the bath
Description
Pk
Cat. No.
DRY-Line 53
1
DRY-Line 115
1
Description
For
Pk
Cat. No.
Shelves
Chrome plated shelf
DL 53/VL 53/IL 53
1
Chrome plated shelf
DL 115/VL 115/IL 115
1
Model
DL 53
DL 115
Capacity (l)
53
115
Convection type
Natural convection
External W×D×H (mm)
620×600×680
820×710×760
Internal W×D×H (mm)
401×330×401
600×400×480
Max. load per shelf (kg)
10
15
Nominal power consumption (W)
800
1000
Shelves supplied (max.)
2 (4)
Temperature fluctuation (time) (°C)
At 150 °C: ±0,4
At 150 °C: ±0,3
Temperature range (°C)
Ambient +5...220
Temperature variation (spatial) (°C)
At 150 °C: ±3,3
At 150 °C: ±3,0
Weight (kg)
41
62
Ovens with natural convection, DRY-Line®
Drying ovens with natural convection used for thermal processes, heated storage and for convection drying
applications with temperatures up to 220 °C. Exhaust duct at the back of the unit with manually adjustable
slide.
Ş Microprocessor controller
Ş Integrated timer can be set for continuous mode or timer operation: 0 to 999 min or 0 to 99,9 h
Ş Over-temperature protection with visual alarm (Class 3,1 safety device)
Ş Digital temperature setting with degree accuracy
Delivery Information:
Supplied with two chrome plated shelves.
1...,233,234,235,236,237,238,239,240,241,242 244,245,246,247,248,249,250,251,252,253,...612
Powered by FlippingBook